Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00652910_s1
          See other VMN1R203 GE Assays ›
          Gene Symbol:
          Vmn1r203
          Gene Name:
          vomeronasal 1 receptor 203
          Gene Aliases:
          V1rh11
          Chromosome Location:
          Chr.13: 22524051 - 22524986 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00652910_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r203
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134236.1 NP_598997.1 1 838 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r203

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068947 NM_134236.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681945 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150548 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068947 NM_134236.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P130663 NM_134236.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28878025 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150548 - -
          Whole Mouse Genome G4122A A_52_P130663 NM_134236.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121861 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150548 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10897394 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10897385 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10897376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28878031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28878003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28878001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28878046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28878030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28878016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28878025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28878038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11100036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28878008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10753393 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403989 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403988 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404002 - -
          Affymetrix Mouse Clariom S Array PSR1300001301.mm.1 NM_134236.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001301.mm.1 NM_134236.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001301.mm.1 TC1300000213.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 359614 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104050717 - -
          Illumina mouse-6 v1.0 expression beadchip 2030551 - -
          Illumina mouse-6 v1.0 expression beadchip 104050717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2420650 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220469 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2420650 - -
          mouse-6 v1.1 (Illumina) GI_21955267-S - -
          mouse-6 v1.1 (Illumina) scl0171270.1_244-S - -
          Illumina Sentrix mouse-6.v1 bead array 2030551 - -
          Illumina Sentrix mouse-6.v1 bead array 104050717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0171270.1_244-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030551 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104050717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220469 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220469 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline