Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653723_s1
          See other OLFR402 GE Assays ›
          Gene Symbol:
          Olfr402
          Gene Name:
          olfactory receptor 402
          Gene Aliases:
          MOR255-4
          Chromosome Location:
          Chr.11: 74155156 - 74156103 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653723_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr402
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146708.1 NP_666919.1 1 451 109
          GenBank mRNA BC116954.1 - 1 492 40126273 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr402

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388268 NM_146708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388268 BC116954.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388268 NM_146708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388268 BC116954.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388268 NM_146708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388268 BC116954.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388268 NM_146708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388268 BC116954.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388268 NM_146708.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388268 BC116954.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388268 NM_146708.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388268 BC116954.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919255 - -
          Whole Mouse Genome G4122A A_51_P388268 NM_146708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388268 BC116954.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388268 NM_146708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388268 BC116954.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105955 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09237902 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09237898 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09195569 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09237904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09237899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09237907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09195576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09237899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09195582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09088227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919238 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10378430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378431 - -
          Affymetrix Mouse Gene 1.1 ST Array 10378429 - -
          Affymetrix Mouse Clariom S Array PSR1100009379.mm.1 TC1100001036.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009380.mm.1 NM_146708.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009380.mm.1 NM_146708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009380.mm.1 TC1100001036.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 691670 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2630433 - -
          Illumina mouse-6 v1.0 expression beadchip 2630433 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002630433 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241786 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241786 - -
          mouse-6 v1.1 (Illumina) scl0258703.1_217-S - -
          Illumina Sentrix mouse-6.v1 bead array 2630433 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258703.1_217-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2630433 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1241786 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241786 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0