Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00654594_s1
          See other OLFR1297 GE Assays ›
          Gene Symbol:
          Olfr1297
          Gene Name:
          olfactory receptor 1297
          Gene Aliases:
          MOR248-4
          Chromosome Location:
          Chr.2: 111621134 - 111626391 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00654594_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1297
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146888.1 NP_667099.1 1 318 153
          GenBank mRNA BC104072.2 - 1 318 40041728 153
          BC104073.2 - 1 318 40041729 153

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1297

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174187 NM_146888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174187 BC104072.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174187 BC104073.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166883 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174187 NM_146888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174187 BC104072.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174187 BC104073.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P207781 NM_146888.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P207781 BC104072.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P207781 BC104073.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526047 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166883 - -
          Whole Mouse Genome G4122A A_51_P207781 NM_146888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P207781 BC104072.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P207781 BC104073.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124792 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01796939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01967205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01464603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01796908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01796920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01796931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526044 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485785 - -
          Affymetrix Mouse Gene 1.1 ST Array 10485784 - -
          Affymetrix Mouse Clariom S Array PSR0200036225.mm.1 NM_146888.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036225.mm.1 NM_146888.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036225.mm.1 TC0200004328.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 471348 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6840685 - -
          Illumina mouse-6 v1.0 expression beadchip 6840685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840685 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2640048 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2640048 - -
          mouse-6 v1.1 (Illumina) scl18876.1.1_95-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18876.1.1_95-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2640048 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2640048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline