Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00658393_m1
          See other GIMAP5 GE Assays ›
          Gene Symbol:
          Gimap5
          Gene Name:
          GTPase, IMAP family member 5
          Gene Aliases:
          D630024P16, E230026N22Rik
          Chromosome Location:
          Chr.6: 48746137 - 48755235 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00658393_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gimap5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175035.5 NP_778200.1 2 - 3 289 76
          GenBank mRNA AB126961.1 - 2 - 3 289 76
          AK052694.1 - 2 - 3 222 76
          BC095995.1 - 2 - 3 201 30470329 76
          BU694320.1 - 2 - 3 247 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003501.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gimap5

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176365 BC095995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176365 AK052694.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176365 NM_175035.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176365 AB126961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176365 BC095995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176365 AK052694.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176365 NM_175035.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176365 AB126961.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369229 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102113 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176365 BC095995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176365 AK052694.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176365 NM_175035.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176365 AB126961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176365 BC095995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176365 AK052694.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176365 NM_175035.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176365 AB126961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176365 BC095995.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176365 AK052694.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176365 NM_175035.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176365 AB126961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176365 BC095995.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176365 AK052694.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176365 NM_175035.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176365 AB126961.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24429136 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01323025 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102113 - -
          Whole Mouse Genome G4122A A_51_P176365 BC095995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176365 AK052694.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176365 NM_175035.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176365 AB126961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176365 BC095995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176365 AK052694.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176365 NM_175035.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P176365 AB126961.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102113 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05362420 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05362415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05788272 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05362419 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05362408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05061522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05362412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05006730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05362420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05061527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05362406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05061526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05138514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05362417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05362419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05362414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05788272 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10538143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538146 - -
          Affymetrix HT MG-430 PM Array Plate 1444181_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10538142 - -
          Mouse Genome 430 2.0 Array 1444181_at - -
          Mouse Genome 430 2.0 Array 1444181_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 317757_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 317757_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600027593.mm.1 TC0600003501.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600027593.mm.1 NM_175035.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600027593.mm.1 NM_175035.5 Refseq
          Mouse Expression Set 430 1444181_at - -
          Mouse Expression Set 430 1444181_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444181_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444181_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 404771 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7050725 - -
          Illumina mouse-6 v1.0 expression beadchip 6420546 - -
          Illumina mouse-6 v1.0 expression beadchip 7050725 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050725 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2643096 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2701332 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2701332 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2643096 - -
          mouse-6 v1.1 (Illumina) scl0317757.1_85-S - -
          mouse-6 v1.1 (Illumina) scl00317757.1_135-S - -
          Illumina Sentrix mouse-6.v1 bead array 6420546 - -
          Illumina Sentrix mouse-6.v1 bead array 7050725 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0317757.1_85-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00317757.1_135-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050725 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420546 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2701332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2643096 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2643096 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2701332 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline