Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00660208_m1
          See other SERPINB9E GE Assays ›
          Gene Symbol:
          Serpinb9e
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade B, member 9e
          Gene Aliases:
          AW540195, NK26, Spi14, ovalbumin
          Chromosome Location:
          Chr.13: 33249610 - 33260846 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00660208_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinb9e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011456.2 NP_035586.1 4 - 5 499 148
          GenBank mRNA AK049429.1 - 4 - 5 502 148
          BC145592.1 - 4 - 5 473 9054158 148
          BC145593.1 - 4 - 5 473 9054159 148
          U96709.2 - 4 - 5 498 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000368.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpinb9e

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459128 BC145592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459128 BC145593.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459128 U96709.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459128 NM_011456.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459128 AK049429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459128 BC145592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459128 BC145593.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459128 U96709.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459128 NM_011456.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459128 AK049429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971430 U96709.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971430 NM_011456.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971430 AK049429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971430 BC050060.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111848 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459128 BC145592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459128 BC145593.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459128 U96709.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459128 NM_011456.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459128 AK049429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459128 BC145592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459128 BC145593.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459128 U96709.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459128 NM_011456.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459128 AK049429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971430 U96709.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971430 NM_011456.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971430 AK049429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971430 BC050060.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28933457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28933453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28933482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28933470 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02660964 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02660961 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07831459 - -
          Whole Mouse Genome G4122A A_51_P459128 BC145592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459128 BC145593.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459128 U96709.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459128 NM_011456.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459128 AK049429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459128 BC145592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459128 BC145593.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459128 U96709.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459128 NM_011456.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459128 AK049429.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07831459 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10962692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28933447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28933542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28933498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28933457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10476271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933453 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404463 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404462 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404482 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404464 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404472 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404479 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404458 - -
          Affymetrix Mouse Clariom S Array PSR1300002212.mm.1 NM_011456.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002212.mm.1 NM_011456.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002212.mm.1 TC1300000368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002213.mm.1 NM_011456.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002213.mm.1 NM_011456.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002213.mm.1 TC1300000368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002210.mm.1 NM_011456.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002210.mm.1 NM_011456.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002210.mm.1 TC1300000368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002215.mm.1 NM_011456.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002215.mm.1 NM_011456.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002215.mm.1 TC1300000368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002211.mm.1 NM_011456.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002211.mm.1 NM_011456.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002211.mm.1 TC1300000368.mm.1 MTA TC
          Mouse Expression Set 430 1418423_s_at - -
          Mouse Expression Set 430 1418423_PM_s_at - -
          Murine Genome U74v2 Set 160972_at - -
          Murine Genome U74v2 Set 161115_r_at - -
          Murine Genome U74v2 Set 161115_PM_r_at - -
          Murine Genome U74v2 Set 160972_PM_at - -
          Mouse Genome 430A 2.0 Array 1418423_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1418423_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 530252 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2480121 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002480121 - -
          mouse-6 v1.1 (Illumina) scl020710.7_139-S - -
          Illumina Sentrix mouse-6.v1 bead array 2480121 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020710.7_139-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2480121 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3006459 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3006459 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2590485 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0