Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00662708_g1
          See other CTS3 GE Assays ›
          Gene Symbol:
          Cts3
          Gene Name:
          cathepsin 3
          Gene Aliases:
          1600000I23Rik
          Chromosome Location:
          Chr.13: 61564630 - 61570162 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00662708_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cts3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026906.3 NP_081182.2 5 - 6 780 149
          NM_054092.2 NP_473433.2 5 - 6 705 149
          GenBank mRNA AK005389.1 - 5 - 6 703 149
          AY034574.1 - 5 - 6 719 149
          BC099388.1 - 5 - 6 674 30786828 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002153.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cts3

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          proteolysis involved in cellular protein catabolic process
          molecular_function
          cysteine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400555 AK005389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400555 AY034574.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400555 BC099388.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400555 NM_026906.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400555 NM_054092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400555 AK005389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400555 AY034574.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400555 BC099388.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400555 NM_026906.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400555 NM_054092.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249644 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260895 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849092 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46613 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400555 AK005389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400555 AY034574.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400555 BC099388.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400555 NM_026906.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400555 NM_054092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400555 AK005389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400555 AY034574.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400555 BC099388.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400555 NM_026906.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400555 NM_054092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400555 AK005389.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400555 AY034574.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400555 BC099388.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400555 NM_026906.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400555 NM_054092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400555 AK005389.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400555 AY034574.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400555 BC099388.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400555 NM_026906.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400555 NM_054092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400555 AK005389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400555 AY034574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400555 BC099388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400555 NM_026906.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400555 NM_054092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400555 AK005389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400555 AY034574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400555 BC099388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400555 NM_026906.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400555 NM_054092.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124991 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10412450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409956 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409957 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409961 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409959 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409950 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409962 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409964 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412458 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409963 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409958 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409960 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409965 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409951 - -
          Affymetrix HT MG-430 PM Array Plate 1453996_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10412454 - -
          Affymetrix Mouse Gene 1.1 ST Array 10409943 - -
          Affymetrix Mouse Gene 1.1 ST Array 10412443 - -
          Affymetrix Mouse Gene 1.1 ST Array 10409955 - -
          Mouse Genome 430 2.0 Array 1453996_PM_a_at - -
          Mouse Genome 430 2.0 Array 1453996_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 117066_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 117066_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300013853.mm.1 NM_026906.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013853.mm.1 NM_026906.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013853.mm.1 NM_054092.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013853.mm.1 NM_054092.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013853.mm.1 TC1300002153.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013859.mm.1 NM_026906.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013859.mm.1 NM_026906.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013859.mm.1 NM_054092.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013859.mm.1 NM_054092.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013859.mm.1 TC1300002153.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013854.mm.1 NM_026906.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013854.mm.1 NM_026906.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013854.mm.1 NM_054092.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013854.mm.1 NM_054092.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013854.mm.1 TC1300002153.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013855.mm.1 NM_026906.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013855.mm.1 NM_026906.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013855.mm.1 NM_054092.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013855.mm.1 NM_054092.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013855.mm.1 TC1300002153.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013856.mm.1 NM_026906.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013856.mm.1 NM_026906.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013856.mm.1 NM_054092.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013856.mm.1 NM_054092.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013856.mm.1 TC1300002153.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013860.mm.1 NM_026906.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013860.mm.1 NM_026906.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013860.mm.1 NM_054092.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013860.mm.1 NM_054092.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013860.mm.1 TC1300002153.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013858.mm.1 NM_026906.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013858.mm.1 NM_026906.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013858.mm.1 TC1300002153.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013857.mm.1 NM_026906.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013857.mm.1 NM_026906.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013857.mm.1 NM_054092.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013857.mm.1 NM_054092.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013857.mm.1 TC1300002153.mm.1 MTA TC
          Mouse Expression Set 430 1453996_a_at - -
          Mouse Expression Set 430 1453996_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1453996_a_at - -
          Mouse Genome 430A 2.0 Array 1453996_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1453996_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1453996_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 761473 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2370397 - -
          Illumina mouse-6 v1.0 expression beadchip 2370397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002370397 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232725 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232725 - -
          mouse-6 v1.1 (Illumina) scl43852.8.1_203-S - -
          Illumina Sentrix mouse-6.v1 bead array 2370397 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43852.8.1_203-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2370397 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2850260 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2850260 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline