Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00728920_s1
          See other OLFR1288 GE Assays ›
          Gene Symbol:
          Olfr1288
          Gene Name:
          olfactory receptor 1288
          Gene Aliases:
          MOR245-9
          Chromosome Location:
          Chr.2: 111478786 - 111479724 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00728920_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1288
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146400.2 NP_666512.2 1 42 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1288

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024270 NM_146400.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814219 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100942 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024270 NM_146400.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P592135 NM_146400.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525463 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100942 - -
          Whole Mouse Genome G4122A A_52_P592135 NM_146400.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103003 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100942 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01965131 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01594409 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01594403 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01594415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02023803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525483 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10474500 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474499 - -
          Affymetrix Mouse Clariom S Array PSR0200012276.mm.1 NM_146400.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012276.mm.1 NM_146400.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012276.mm.1 TC0200001648.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1450593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2685373 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2685373 - -
          mouse-6 v1.1 (Illumina) scl20493.1.1_136-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20493.1.1_136-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2685373 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2685373 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline