Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729356_s1
          See other OLFR1241 GE Assays ›
          Gene Symbol:
          Olfr1241
          Gene Name:
          olfactory receptor 1241
          Gene Aliases:
          MOR231-14
          Chromosome Location:
          Chr.2: 89482189 - 89483133 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729356_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1241
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146455.1 NP_666666.1 1 560 140

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1241

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P126089 NM_146455.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P126089 NM_146455.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P126089 NM_146455.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P126089 NM_146455.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P126089 NM_146455.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P126089 NM_146455.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126089 NM_146455.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126089 NM_146455.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01219468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01721147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01219469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413945 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484825 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484824 - -
          Affymetrix Mouse Clariom S Array PSR0200034058.mm.1 NM_146455.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034058.mm.1 NM_146455.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034058.mm.1 TC0200004060.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3390497 - -
          Illumina mouse-6 v1.0 expression beadchip 3390497 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003390497 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232688 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232688 - -
          mouse-6 v1.1 (Illumina) scl19025.1.346_74-S - -
          Illumina Sentrix mouse-6.v1 bead array 3390497 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19025.1.346_74-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3390497 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2991761 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2991761 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline