Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729373_s1
          See other OLFR282 GE Assays ›
          Gene Symbol:
          Olfr282
          Gene Name:
          olfactory receptor 282
          Gene Aliases:
          MOR160-2
          Chromosome Location:
          Chr.15: 98437470 - 98438397 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729373_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr282
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146457.2 NP_666668.2 1 448 78
          GenBank mRNA BC128027.1 - 1 451 78

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr282

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474567 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474565 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006437 NM_146457.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006437 BC128027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006436 NM_146457.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103223 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006436 NM_146457.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006437 NM_146457.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006437 BC128027.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320751 NM_146457.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320751 BC128027.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474564 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103223 - -
          Whole Mouse Genome G4122A A_51_P320751 NM_146457.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320751 BC128027.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101592 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103223 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12459308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12459311 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474565 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30474567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12459311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12244691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12044845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12244697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12459306 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10426593 - -
          Affymetrix Mouse Gene 1.1 ST Array 10426592 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 664493 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2100440 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002100440 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2752262 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2752262 - -
          mouse-6 v1.1 (Illumina) scl0258449.1_100-S - -
          Illumina Sentrix mouse-6.v1 bead array 2100440 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258449.1_100-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2100440 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2752262 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2752262 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline