Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729479_s1
          See other OLFR1384 GE Assays ›
          Gene Symbol:
          Olfr1384
          Gene Name:
          olfactory receptor 1384
          Gene Aliases:
          MOR256-23
          Chromosome Location:
          Chr.11: 49513618 - 49514651 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729479_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1384
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146472.2 NP_666683.1 1 274 99
          GenBank mRNA BC119159.1 - 1 274 8733908 99
          BC120667.1 - 1 274 40129590 99

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1384

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347942 NM_146472.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347942 BC119159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347942 BC120667.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347942 NM_146472.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347942 BC119159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347942 BC120667.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120150 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347942 NM_146472.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347942 BC119159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347942 BC120667.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347942 NM_146472.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347942 BC119159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347942 BC120667.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P347942 NM_146472.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P347942 BC119159.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P347942 BC120667.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P347942 NM_146472.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P347942 BC119159.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P347942 BC120667.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783263 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120150 - -
          Whole Mouse Genome G4122A A_52_P347942 NM_146472.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347942 BC119159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347942 BC120667.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347942 NM_146472.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347942 BC119159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347942 BC120667.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119855 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130551 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120150 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09163471 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09163496 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09163492 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09163467 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09163476 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09163494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09163493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09163496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09163459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09091695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09091692 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375569 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375568 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375567 - -
          Affymetrix Mouse Clariom S Array PSR1100003911.mm.1 NM_146472.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003911.mm.1 NM_146472.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003911.mm.1 TC1100000514.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 439174 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5220161 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220161 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2670532 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2670532 - -
          mouse-6 v1.1 (Illumina) scl41626.1.1_322-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220161 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41626.1.1_322-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220161 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2670532 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2670532 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41626.1.1_322-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline