Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730406_s1
          See other OLFR435 GE Assays ›
          Gene Symbol:
          Olfr435
          Gene Name:
          olfactory receptor 435
          Gene Aliases:
          MOR261-5
          Chromosome Location:
          Chr.6: 43201646 - 43202587 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730406_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr435
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146653.1 NP_666864.1 1 152 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr435

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P90463 NM_146653.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P90463 NM_146653.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152122 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P90463 NM_146653.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P90463 NM_146653.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P90463 NM_146653.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P90463 NM_146653.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24402030 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152122 - -
          Whole Mouse Genome G4122A A_52_P90463 NM_146653.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P90463 NM_146653.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131967 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152122 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05627036 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05627056 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05627057 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05627051 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05627060 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05627035 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05627049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24402001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05627051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05627034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24402045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401987 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537827 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537826 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 60280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000060280 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1250611 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250611 - -
          mouse-6 v1.1 (Illumina) scl30117.1.1_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 60280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30117.1.1_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 60280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2907173 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1250611 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1250611 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2907173 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline