Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00731448_s1
          See other SLFN10-PS GE Assays ›
          Gene Symbol:
          Slfn10-ps
          Gene Name:
          schlafen 10, pseudogene
          Gene Aliases:
          Slfn10, mFLJ00257
          Chromosome Location:
          Chr.11: 83028123 - 83040533 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00731448_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slfn10-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_073523.1 2 670 98
          NR_073524.1 2 545 98
          GenBank mRNA AK144698.1 - 2 546 98
          AK220241.1 - 2 670 98
          AY261802.1 - 2 545 98
          AY261803.1 - 2 514 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135200 AY261803.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135200 BP765349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135200 AK220241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135200 AY261802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135200 AK144698.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284341 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 117485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129803 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159007 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135200 AY261803.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135200 BP765349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135200 AK220241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135200 AY261802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135200 AK144698.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969747 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159007 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07420006 - -
          Whole Mouse Genome G4122A A_51_P504991 BC152549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504991 NM_001167743.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504991 NM_181545.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504991 AK150875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504991 AY261803.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130199 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09505159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09543837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09117886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09263207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09644867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09695497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969736 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10389155 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389154 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389158 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389156 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389157 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389160 - -
          Affymetrix HT MG-430 PM Array Plate 1444350_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10389151 - -
          Mouse Genome 430 2.0 Array 1444350_PM_at - -
          Mouse Genome 430 2.0 Array 1444350_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237887_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237887_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100031023.mm.1 NR_073523.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031023.mm.1 NR_073524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031023.mm.1 TC1100003385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031022.mm.1 NR_073523.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031022.mm.1 NR_073524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031022.mm.1 TC1100003385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031025.mm.1 NR_073523.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031025.mm.1 NR_073524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031025.mm.1 TC1100003385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031024.mm.1 NR_073523.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031024.mm.1 NR_073524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031024.mm.1 TC1100003385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031026.mm.1 NR_073523.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031026.mm.1 NR_073524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031026.mm.1 TC1100003385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031029.mm.1 NR_073524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031029.mm.1 TC1100003385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031028.mm.1 NR_073523.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031028.mm.1 TC1100003385.mm.1 MTA TC
          Mouse Expression Set 430 1444350_at - -
          Mouse Expression Set 430 1444350_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444350_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444350_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 322234 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2510068 - -
          Illumina mouse-6 v1.0 expression beadchip 2510068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002510068 - -
          mouse-6 v1.1 (Illumina) scl39827.10.1_105-S - -
          Illumina Sentrix mouse-6.v1 bead array 2510068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39827.10.1_105-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2510068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3060838 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3138280 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3060838 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3138280 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2735689 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline