Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732688_s1
          See other OLFR1434 GE Assays ›
          Gene Symbol:
          Olfr1434
          Gene Name:
          olfactory receptor 1434
          Gene Aliases:
          MOR214-4, Olfr1433, Olfr1433-ps1
          Chromosome Location:
          Chr.19: 12279854 - 12283988 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732688_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1434
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146685.3 NP_666896.2 2 813 127
          GenBank mRNA BC132136.1 - 1 741 40130413 127
          BC138096.1 - 1 741 8861115 127
          BC147691.1 - 1 742 9056664 127
          BC147692.1 - 1 742 9056665 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000241.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1434

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994837 BC147692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994837 BC132136.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994837 BC147691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994837 NM_207626.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994837 BC138096.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108435 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108435 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994837 BC147692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994837 BC132136.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994837 BC147691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994837 NM_207626.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994837 BC138096.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P524988 BC147692.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P524988 BC132136.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P524988 BC147691.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P524988 NM_207626.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P524988 BC138096.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960190 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108435 - -
          Whole Mouse Genome G4122A A_52_P524988 BC147692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P524988 BC132136.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P524988 BC147691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P524988 NM_207626.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P524988 BC138096.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133509 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108435 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14184910 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198986 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14184908 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14199020 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14223281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14505740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14505751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14199002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14442262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14184904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14442264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960171 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461712 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461711 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461711 - -
          Affymetrix Mouse Clariom S Array PSR1900002889.mm.1 TC1900000241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002884.mm.1 NM_146685.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002884.mm.1 NM_146685.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002884.mm.1 TC1900000241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002886.mm.1 TC1900000241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002888.mm.1 NM_146685.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002888.mm.1 NM_146685.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002888.mm.1 TC1900000241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002890.mm.1 TC1900000241.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3870609 - -
          Illumina mouse-6 v1.0 expression beadchip 3870609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870609 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2639249 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2639249 - -
          mouse-6 v1.1 (Illumina) scl00258680.1_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 3870609 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258680.1_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870609 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2944076 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2944076 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2639249 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline