Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00776302_g1
          See other KLK1B8 GE Assays ›
          Gene Symbol:
          Klk1b8
          Gene Name:
          kallikrein 1-related peptidase b8
          Gene Aliases:
          Klk8, Nrpn, TADG14, mGK-8
          Chromosome Location:
          Chr.7: 43950672 - 43954938 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00776302_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klk1b8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008457.2 NP_032483.1 4 - 5 645 153
          GenBank mRNA BC031798.1 - 4 - 5 650 4167600 153
          X03994.1 - 4 - 5 644 153

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000783.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klk1b8

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155503 X03994.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155503 NM_008457.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155503 BC031798.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155503 X03994.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155503 NM_008457.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155503 BC031798.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155503 X03994.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155503 NM_008457.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155503 BC031798.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155503 X03994.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155503 NM_008457.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155503 BC031798.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25084899 - -
          Whole Mouse Genome G4122A A_51_P155503 X03994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155503 NM_008457.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155503 BC031798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155503 X03994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155503 NM_008457.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155503 BC031798.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119831 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129979 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06564006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25084887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06017783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06318354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06613419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06613417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06613405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06017780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06318355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06613406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25084878 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552561 - -
          Affymetrix HT MG-430 PM Array Plate 1449463_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552557 - -
          Mouse Genome 430 2.0 Array 1449463_at - -
          Mouse Genome 430 2.0 Array 1449463_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16624_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16624_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700006353.mm.1 TC0700000783.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006353.mm.1 NM_008457.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006354.mm.1 TC0700000783.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006354.mm.1 NM_008457.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006350.mm.1 TC0700000783.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006350.mm.1 NM_008457.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006352.mm.1 TC0700000783.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006352.mm.1 NM_008457.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006351.mm.1 TC0700000783.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006351.mm.1 NM_008457.3 Refseq
          Mouse Expression Set 430 1449463_PM_at - -
          Mouse Expression Set 430 1449463_at - -
          Mouse Genome 430A 2.0 Array 1449463_at - -
          Mouse Genome 430A 2.0 Array 1449463_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449463_at - -
          Affymetrix HT Mouse Genome 430A Array 1449463_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 767496 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 60035 - -
          Illumina mouse-6 v1.0 expression beadchip 60035 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000060035 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216962 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216962 - -
          mouse-6 v1.1 (Illumina) scl32735.4_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 60035 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32735.4_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 60035 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216962 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216962 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline