Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00778687_s1
          See other APH1A GE Assays ›
          Gene Symbol:
          Aph1a
          Gene Name:
          anterior pharynx defective 1a homolog (C. elegans)
          Gene Aliases:
          6530402N02Rik, APH-1a
          Chromosome Location:
          Chr.3: 95893907 - 95898592 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00778687_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aph1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146104.3 NP_666216.1 6 2855 141
          NM_146134.2 NP_666246.1 7 2522 141
          GenBank mRNA AK148400.1 - 6 2808 141
          BC012406.1 - 6 2780 3483214 141

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Mouse Alzheimer's Disease Inventoried Mouse_Alzheimer Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aph1a

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          metanephros development
          proteolysis
          membrane protein ectodomain proteolysis
          Notch signaling pathway
          Notch receptor processing
          protein processing
          membrane protein intracellular domain proteolysis
          amyloid precursor protein catabolic process
          positive regulation of catalytic activity
          endopeptidase activity
          protein binding
          peptidase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22346232 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22346233 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401964 BC012406.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015044 AK216088.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015044 BC012406.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015044 BC024111.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015044 BC057865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015044 AK148400.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694739 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741710 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155265 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015044 AK216088.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015044 BC012406.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015044 BC024111.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015044 BC057865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015044 AK148400.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401964 BC012406.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P217195 BC012406.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P217195 BC024111.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P217195 BC057865.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P217195 AK148400.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P217195 AK152334.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P9716 BC012406.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P9716 BC024111.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P9716 BC057865.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P9716 AK148400.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P9716 AK152334.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22346247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22346233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22346243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22346242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22346239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22346248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22346236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22346241 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155265 - -
          Whole Mouse Genome G4122A A_52_P217195 BC012406.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P217195 BC024111.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P217195 BC057865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P217195 AK148400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P217195 AK152334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P9716 BC012406.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P9716 BC024111.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P9716 BC057865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P9716 AK148400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P9716 AK152334.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103530 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106908 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155265 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02295301 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22346232 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22346233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22346218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02295279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02295269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02295298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22346233 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10494317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359429 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494319 - -
          Affymetrix HT MG-430 PM Array Plate 1424980_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1424979_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451554_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10500283 - -
          Affymetrix Mouse Gene 1.1 ST Array 10359428 - -
          Affymetrix Mouse Gene 1.1 ST Array 10494312 - -
          Mouse Genome 430 2.0 Array 1451554_PM_a_at - -
          Mouse Genome 430 2.0 Array 1424979_PM_at - -
          Mouse Genome 430 2.0 Array 1424979_at - -
          Mouse Genome 430 2.0 Array 1424980_PM_s_at - -
          Mouse Genome 430 2.0 Array 1451554_a_at - -
          Mouse Genome 430 2.0 Array 1424980_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226548_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226548_at - -
          Affymetrix Mouse Clariom S Array PSR0300007294.mm.1 NM_146104.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007294.mm.1 NM_146134.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007294.mm.1 TC0300000914.mm.1 MTA TC
          Mouse Expression Set 430 1451554_a_at - -
          Mouse Expression Set 430 1424980_PM_s_at - -
          Mouse Expression Set 430 1424979_at - -
          Mouse Expression Set 430 1451554_PM_a_at - -
          Mouse Expression Set 430 1424980_s_at - -
          Mouse Expression Set 430 1424979_PM_at - -
          Murine Genome U74v2 Set 98078_at - -
          Murine Genome U74v2 Set 98078_PM_at - -
          Mouse Genome 430A 2.0 Array 1424979_PM_at - -
          Mouse Genome 430A 2.0 Array 1424979_at - -
          Mouse Genome 430A 2.0 Array 1424980_s_at - -
          Mouse Genome 430A 2.0 Array 1451554_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1451554_a_at - -
          Mouse Genome 430A 2.0 Array 1424980_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424979_at - -
          Affymetrix HT Mouse Genome 430A Array 1451554_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451554_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424979_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424980_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424980_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6450711 - -
          Illumina mouse-6 v1.0 expression beadchip 4060273 - -
          Illumina mouse-6 v1.0 expression beadchip 4670465 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670465 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2750628 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232821 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2588759 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2709412 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217553 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232821 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2750628 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2709412 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2588759 - -
          mouse-6 v1.1 (Illumina) scl22874.5.1_2-S - -
          mouse-6 v1.1 (Illumina) scl0226548.6_115-S - -
          mouse-6 v1.1 (Illumina) scl00226548.1_110-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450711 - -
          Illumina Sentrix mouse-6.v1 bead array 4670465 - -
          Illumina Sentrix mouse-6.v1 bead array 4060273 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22874.5.1_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0226548.6_115-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00226548.1_110-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450711 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060273 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670465 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232821 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2750628 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232821 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2750628 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl22874.5.1_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0