Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00780670_s1
          See other DPH3 GE Assays ›
          Gene Symbol:
          Dph3
          Gene Name:
          diphthamine biosynthesis 3
          Gene Aliases:
          DELGIP1, DelgipP1, Desr1, KTI11, Zcsl2
          Chromosome Location:
          Chr.14: 32080517 - 32085692 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00780670_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dph3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047433.2 NP_001040898.1 2 2884 99
          NM_001284346.1 NP_001271275.1 4 1060 99
          NM_172254.4 NP_758458.1 3 2959 99
          NR_104303.1 3 2817 99
          NR_104304.1 3 3085 99
          NR_104305.1 2 2981 99
          GenBank mRNA AK048097.1 - 3 2849 99
          BC039954.1 - 4 971 4193739 99

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dph3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein secretion
          positive regulation of binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29515525 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29515529 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29515523 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29515524 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29515526 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063495 BC039954.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063495 NM_172254.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063495 NM_001047433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080318 AK048097.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080318 NM_172254.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080318 NM_001047433.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353844 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817904 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41940 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101699 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080318 AK048097.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080318 NM_172254.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080318 NM_001047433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063495 BC039954.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063495 NM_172254.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063495 NM_001047433.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328789 AK048097.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328789 NM_172254.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328789 AK017764.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328789 NM_001047433.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P291745 AK048097.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P291745 BC039954.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P291745 NM_172254.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P291745 NM_001047433.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515487 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101699 - -
          Whole Mouse Genome G4122A A_51_P328789 AK048097.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328789 NM_172254.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328789 AK017764.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328789 NM_001047433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P521710 AK048097.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P521710 BC029910.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P521710 NM_172254.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P521710 AK011448.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P521710 AK017764.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P291745 AK048097.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P291745 BC039954.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P291745 NM_172254.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P291745 NM_001047433.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105294 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101699 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11806692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11806673 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11806682 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29515526 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29515523 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29515524 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29515525 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29515529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11490515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11158318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11490513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11463237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11638429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11765335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11158319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11463231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11638431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515513 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10418803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418801 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418799 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418798 - -
          Affymetrix HT MG-430 PM Array Plate 1431200_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1433921_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1455268_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10418796 - -
          Mouse Genome 430 2.0 Array 1431200_a_at - -
          Mouse Genome 430 2.0 Array 1455268_PM_at - -
          Mouse Genome 430 2.0 Array 1433921_s_at - -
          Mouse Genome 430 2.0 Array 1433921_PM_s_at - -
          Mouse Genome 430 2.0 Array 1455268_at - -
          Mouse Genome 430 2.0 Array 1431200_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 105638_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 105638_at - -
          Affymetrix Mouse Clariom S Array PSR1400013136.mm.1 NM_001047433.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013136.mm.1 NM_001284346.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013136.mm.1 NM_172254.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013136.mm.1 NR_104303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013136.mm.1 NR_104304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013136.mm.1 NR_104305.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013136.mm.1 TC1400001695.mm.1 MTA TC
          Mouse Expression Set 430 1455268_PM_at - -
          Mouse Expression Set 430 1431200_PM_a_at - -
          Mouse Expression Set 430 1431200_a_at - -
          Mouse Expression Set 430 1455268_at - -
          Mouse Expression Set 430 1433921_PM_s_at - -
          Mouse Expression Set 430 1433921_s_at - -
          Murine Genome U74v2 Set 165989_f_at - -
          Murine Genome U74v2 Set 170947_PM_at - -
          Murine Genome U74v2 Set 170947_at - -
          Murine Genome U74v2 Set 165989_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1433921_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1433921_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1431200_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1455268_at - -
          Affymetrix HT Mouse Genome 430B Array 1455268_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431200_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 870731 - -
          MouseWG-6 v2 Expression BeadChip 6980114 - -
          Illumina mouse-6 v1.0 expression beadchip 6980114 - -
          Illumina mouse-6 v1.0 expression beadchip 870731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000870731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2717172 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2704641 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2704641 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2717172 - -
          mouse-6 v1.1 (Illumina) scl45748.3_30-S - -
          mouse-6 v1.1 (Illumina) scl000359.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980114 - -
          Illumina Sentrix mouse-6.v1 bead array 870731 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000359.1_0-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45748.3_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980114 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 870731 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2704641 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3134412 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2717172 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2704641 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2717172 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3134412 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl45748.3_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000359.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline