Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00812903_m1
          See other RNF20 GE Assays ›
          Gene Symbol:
          Rnf20
          Gene Name:
          ring finger protein 20
          Gene Aliases:
          4833430L21Rik, AW540162, C79397, mKIAA4116
          Chromosome Location:
          Chr.4: 49632011 - 49656886 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00812903_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163263.1 NP_001156735.1 2 - 3 610 74
          NM_182999.2 NP_892044.1 3 - 4 457 74
          GenBank mRNA AK048862.1 - 2 - 3 569 74
          AK051278.1 - 2 - 3 569 74
          AK053783.1 - 3 - 4 445 74
          AK082529.1 - 2 - 3 513 74
          AK085436.1 - 2 - 3 588 74
          AK087860.1 - 2 - 3 534 74
          AK139883.1 - 3 - 4 435 74
          AK220492.1 - 2 - 3 526 74
          BC052482.1 - 3 - 4 396 6308280 74
          CJ052397.1 - 3 - 4 438 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000555.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf20

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          regulation of transcription, DNA-templated
          ubiquitin-dependent protein catabolic process
          histone monoubiquitination
          chromatin modification
          histone ubiquitination
          negative regulation of cell migration
          positive regulation of histone methylation
          histone H2B ubiquitination
          positive regulation of transcription, DNA-templated
          negative regulation of mRNA polyadenylation
          positive regulation of histone H2B ubiquitination
          p53 binding
          chromatin binding
          transcription coactivator activity
          mRNA 3'-UTR binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          ubiquitin protein ligase binding
          histone binding
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400004062.mm.1 NM_001163263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004062.mm.1 NM_001163263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004062.mm.1 NM_182999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004062.mm.1 NM_182999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004062.mm.1 TC0400000555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004059.mm.1 NM_001163263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004059.mm.1 NM_001163263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004059.mm.1 TC0400000555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004054.mm.1 NM_001163263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004054.mm.1 NM_001163263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004054.mm.1 NM_182999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004054.mm.1 NM_182999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004054.mm.1 TC0400000555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004060.mm.1 NM_001163263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004060.mm.1 NM_001163263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004060.mm.1 NM_182999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004060.mm.1 NM_182999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004060.mm.1 TC0400000555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004061.mm.1 NM_001163263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004061.mm.1 NM_001163263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004061.mm.1 NM_182999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004061.mm.1 NM_182999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004061.mm.1 TC0400000555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004058.mm.1 NM_001163263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004058.mm.1 NM_001163263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004058.mm.1 NM_182999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004058.mm.1 NM_182999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004058.mm.1 TC0400000555.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004055.mm.1 NM_001163263.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004055.mm.1 NM_001163263.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004055.mm.1 NM_182999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004055.mm.1 NM_182999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004055.mm.1 TC0400000555.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 879304 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline