Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00834759_gH
          See other KLK1B27 GE Assays ›
          Gene Symbol:
          Klk1b27
          Gene Name:
          kallikrein 1-related peptidase b27
          Gene Aliases:
          Gk27, Klk21l, Klk27, mGK-27
          Chromosome Location:
          Chr.7: 44052290 - 44056712 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00834759_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klk1b27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020268.3 NP_064664.1 4 - 5 686 188
          GenBank mRNA AB039275.1 - 4 - 5 688 188
          BC132090.1 - 4 - 5 663 40130367 188
          BC132665.1 - 4 - 5 663 40130942 188

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000788.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klk1b27

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419656 BC132665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419656 NM_020268.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419656 AB039275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419656 BC132090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419656 BC132665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419656 NM_020268.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419656 AB039275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P419656 BC132090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P419656 BC132665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P419656 NM_020268.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P419656 AB039275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P419656 BC132090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P419656 BC132665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P419656 NM_020268.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P419656 AB039275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P419656 BC132090.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419656 BC132665.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419656 NM_020268.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419656 AB039275.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419656 BC132090.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419656 BC132665.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419656 NM_020268.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419656 AB039275.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419656 BC132090.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085446 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06178996 - -
          Whole Mouse Genome G4122A A_51_P419656 BC132665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419656 NM_020268.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419656 AB039275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419656 BC132090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419656 BC132665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419656 NM_020268.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419656 AB039275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419656 BC132090.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139844 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06481921 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06481919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06398628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06084530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06520346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06553039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06481921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085422 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552582 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552583 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552590 - -
          Affymetrix HT MG-430 PM Array Plate 1421587_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552587 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552581 - -
          Mouse Genome 430 2.0 Array 1421587_PM_at - -
          Mouse Genome 430 2.0 Array 1421587_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16619_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16619_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700006391.mm.1 TC0700000788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006391.mm.1 NM_020268.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006391.mm.1 NM_020268.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006393.mm.1 TC0700000788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006393.mm.1 NM_020268.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006393.mm.1 NM_020268.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006394.mm.1 TC0700000788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006394.mm.1 NM_020268.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006394.mm.1 NM_020268.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006392.mm.1 TC0700000788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006392.mm.1 NM_020268.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006392.mm.1 NM_020268.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006395.mm.1 TC0700000788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006395.mm.1 NM_020268.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006395.mm.1 NM_020268.3 Refseq
          Mouse Expression Set 430 1421587_PM_at - -
          Mouse Expression Set 430 1421587_at - -
          Murine Genome U74v2 Set 162456_f_at - -
          Murine Genome U74v2 Set 162456_PM_f_at - -
          Murine Genome U74v2 Set 170458_r_at - -
          Murine Genome U74v2 Set 169088_at - -
          Murine Genome U74v2 Set 169088_PM_at - -
          Murine Genome U74v2 Set 170458_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1421587_PM_at - -
          Mouse Genome 430A 2.0 Array 1421587_at - -
          Affymetrix HT Mouse Genome 430A Array 1421587_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421587_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 556228 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2810008 - -
          Illumina mouse-6 v1.0 expression beadchip 5900632 - -
          Illumina mouse-6 v1.0 expression beadchip 103800270 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002810008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005900632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252131 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252131 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237352 - -
          mouse-6 v1.1 (Illumina) scl0016619.1_79-S - -
          mouse-6 v1.1 (Illumina) GI_12313876-S - -
          mouse-6 v1.1 (Illumina) scl016619.3_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 103800270 - -
          Illumina Sentrix mouse-6.v1 bead array 2810008 - -
          Illumina Sentrix mouse-6.v1 bead array 5900632 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl016619.3_29-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0016619.1_79-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5900632 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103800270 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2810008 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3009447 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252131 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3009447 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline