Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00835868_s1
          See other VMN1R42 GE Assays ›
          Gene Symbol:
          Vmn1r42
          Gene Name:
          vomeronasal 1 receptor 42
          Gene Aliases:
          V1ra1, V1ra3, V1ra6
          Chromosome Location:
          Chr.6: 89844518 - 89845615 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00835868_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r42
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053221.2 NP_444451.2 1 732 159
          GenBank mRNA BC125372.1 - 1 732 40129887 159

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r42

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of chemical stimulus
          response to pheromone
          signal transducer activity
          receptor activity
          G-protein coupled receptor activity
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24630038 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24630039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24630035 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24630036 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243094 BC125372.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243094 NM_053221.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243094 BC125372.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P243094 NM_053221.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243094 BC125372.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243094 NM_053221.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243094 BC125372.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P243094 NM_053221.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243094 BC125372.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243094 NM_053221.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243094 BC125372.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243094 NM_053221.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630060 - -
          Whole Mouse Genome G4122A A_51_P243094 BC125372.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243094 NM_053221.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243094 BC125372.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243094 NM_053221.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106915 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131729 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05819500 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05819492 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05819484 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24630036 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24630039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24630035 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24630038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05067014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05819484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05067025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05865316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05057714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05819496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05711824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630060 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10546232 - -
          Affymetrix HT MG-430 PM Array Plate 1422369_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10546231 - -
          Mouse Genome 430 2.0 Array 1422369_at - -
          Mouse Genome 430 2.0 Array 1422369_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 113848_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 113848_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600021046.mm.1 TC0600002703.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600021046.mm.1 NM_053221.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600021046.mm.1 NM_053221.2 Refseq
          Mouse Expression Set 430 1422369_PM_at - -
          Mouse Expression Set 430 1422369_at - -
          Mouse Genome 430A 2.0 Array 1422369_PM_at - -
          Mouse Genome 430A 2.0 Array 1422369_at - -
          Affymetrix HT Mouse Genome 430A Array 1422369_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422369_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 305590 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2120014 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120014 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248443 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248443 - -
          mouse-6 v1.1 (Illumina) scl839.1.1_80-S - -
          Illumina Sentrix mouse-6.v1 bead array 2120014 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl839.1.1_80-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120014 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248443 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248443 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline