Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00835911_s1
          See other VMN1R11 GE Assays ›
          Gene Symbol:
          Vmn1r11
          Gene Name:
          vomeronasal 1 receptor 11
          Gene Aliases:
          V1rc3
          Chromosome Location:
          Chr.6: 57137353 - 57138252 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00835911_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053233.2 NP_444463.2 1 833 122
          GenBank mRNA BC104398.1 - 1 833 40050459 122
          BC104399.1 - 1 833 40050460 122

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r11

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          sensory perception of chemical stimulus
          receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P278606 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127701 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P278606 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P278606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478093 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127701 - -
          Whole Mouse Genome G4122A A_51_P278606 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05010889 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05010899 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05010884 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05010886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05835796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05010880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24478101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05399680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05399682 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10538574 - -
          Affymetrix HT MG-430 PM Array Plate 1422391_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10538573 - -
          Mouse Genome 430 2.0 Array 1422391_PM_at - -
          Mouse Genome 430 2.0 Array 1422391_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 113860_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 113860_at - -
          Mouse Expression Set 430 1422391_PM_at - -
          Mouse Expression Set 430 1422391_at - -
          Mouse Genome 430A 2.0 Array 1422391_at - -
          Mouse Genome 430A 2.0 Array 1422391_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422391_at - -
          Affymetrix HT Mouse Genome 430A Array 1422391_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 713538 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4670204 - -
          Illumina mouse-6 v1.0 expression beadchip 4670204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2480013 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2480013 - -
          mouse-6 v1.1 (Illumina) scl1658.1.1_187-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl1658.1.1_187-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2895819 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2895819 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline