Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836129_s1
          See other VMN1R30 GE Assays ›
          Gene Symbol:
          Vmn1r30
          Gene Name:
          vomeronasal 1 receptor 30
          Gene Aliases:
          V1rc22, V1rc9
          Chromosome Location:
          Chr.6: 58434848 - 58521728 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836129_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r30
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134177.1 NP_598938.1 1 163 187
          GenBank mRNA BC138075.1 - 1 212 8861094 187
          BC138078.1 - 1 212 8861097 187

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002372.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r30

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of chemical stimulus
          response to pheromone
          receptor activity
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2034610 BC138078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2034610 NM_134177.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2034610 BC138075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019978 BC138078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019978 NM_134177.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019978 BC138075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019978 BC138078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019978 NM_134177.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019978 BC138075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2034610 BC138078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2034610 NM_134177.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2034610 BC138075.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24481830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24481814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24481826 - -
          Whole Mouse Genome G4122A A_52_P492799 BC138078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P492799 NM_134177.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P492799 BC138075.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24481846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24481830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24481844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05054208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05323240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05054209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05354466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481826 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545040 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545039 - -
          Affymetrix Mouse Gene 1.1 ST Array 10545038 - -
          Mouse Genome 430 2.0 Array 1422395_PM_at - -
          Mouse Genome 430 2.0 Array 1422395_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 171195_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 171195_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600018402.mm.1 NM_134177.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018402.mm.1 NM_134177.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018402.mm.1 TC0600002372.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018401.mm.1 TC0600002372.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018403.mm.1 TC0600002372.mm.1 MTA TC
          Mouse Expression Set 430 1422395_at - -
          Mouse Expression Set 430 1422395_PM_at - -
          Mouse Genome 430A 2.0 Array 1422395_at - -
          Mouse Genome 430A 2.0 Array 1422395_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 490370 - -
          ABI Mouse Genome Survey Microarray 762275 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2850292 - -
          MouseWG-6 v2 Expression BeadChip 3390504 - -
          Illumina mouse-6 v1.0 expression beadchip 2850292 - -
          Illumina mouse-6 v1.0 expression beadchip 105570435 - -
          Illumina mouse-6 v1.0 expression beadchip 3390504 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003390504 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850292 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2484864 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2478278 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2587700 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2484864 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2478278 - -
          mouse-6 v1.1 (Illumina) scl0001230.1_0-S - -
          mouse-6 v1.1 (Illumina) GI_21717682-I - -
          mouse-6 v1.1 (Illumina) scl0171195.1_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 3390504 - -
          Illumina Sentrix mouse-6.v1 bead array 2850292 - -
          Illumina Sentrix mouse-6.v1 bead array 105570435 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001230.1_0-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0171195.1_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850292 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105570435 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3390504 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2484864 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2899010 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2899011 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2484864 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2899011 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2587700 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2899010 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline