Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838831_g1
          See other ISG20L2 GE Assays ›
          Gene Symbol:
          Isg20l2
          Gene Name:
          interferon stimulated exonuclease gene 20-like 2
          Gene Aliases:
          4930429N24, C80587
          Chromosome Location:
          Chr.3: 87930314 - 87940686 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838831_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Isg20l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177663.4 NP_808331.1 2 - 3 1240 75
          GenBank mRNA AK053778.1 - 2 - 3 1241 75
          AK076747.1 - 2 - 3 963 75
          AK155979.1 - 2 - 3 1244 75
          BC098326.1 - 5720443 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Isg20l2

          Panther Classification:

          Molecular Function -

          exoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          ribosome biogenesis
          nucleic acid binding
          nuclease activity
          exonuclease activity
          hydrolase activity
          phosphoric ester hydrolase activity
          T/G mismatch-specific endonuclease activity
          poly(A) RNA binding
          retroviral 3' processing activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310780 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310781 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028758 BC098326.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028758 AK155979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028758 NM_177663.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028758 AK053778.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P28792 BC098326.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P28792 NM_177663.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P28792 AK053778.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P28792 BC098326.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P28792 NM_177663.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P28792 AK053778.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2054653 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 934788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 617243 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178996 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140626 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028758 BC098326.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028758 AK155979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028758 NM_177663.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028758 AK053778.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P28792 BC098326.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P28792 NM_177663.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P28792 AK053778.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P28792 BC098326.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P28792 NM_177663.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P28792 AK053778.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2054653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310795 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178996 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140626 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00693959 - -
          Whole Mouse Genome G4122A A_52_P572456 BC098326.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P572456 AK076747.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P572456 AK155979.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P572456 AK178428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P572456 NM_177663.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P28792 BC098326.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P28792 NM_177663.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P28792 AK053778.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P28792 BC098326.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P28792 NM_177663.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P28792 AK053778.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101789 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178996 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140626 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02367975 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02367976 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310780 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02930084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03061038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310803 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493104 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493105 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493106 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493107 - -
          Affymetrix HT MG-430 PM Array Plate 1433960_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493103 - -
          Mouse Genome 430 2.0 Array 1433960_at - -
          Mouse Genome 430 2.0 Array 1433960_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 229504_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 229504_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300004788.mm.1 NM_177663.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004788.mm.1 NM_177663.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004788.mm.1 TC0300000709.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004785.mm.1 NM_177663.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004785.mm.1 NM_177663.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004785.mm.1 TC0300000709.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004786.mm.1 NM_177663.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004786.mm.1 NM_177663.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004786.mm.1 TC0300000709.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004787.mm.1 NM_177663.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004787.mm.1 NM_177663.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004787.mm.1 TC0300000709.mm.1 MTA TC
          Mouse Expression Set 430 1433960_at - -
          Mouse Expression Set 430 1433960_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433960_at - -
          Affymetrix HT Mouse Genome 430B Array 1433960_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 872493 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4850008 - -
          Illumina mouse-6 v1.0 expression beadchip 4850008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850008 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226070 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226070 - -
          mouse-6 v1.1 (Illumina) scl23014.5.1_141-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850008 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23014.5.1_141-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850008 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2996667 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2996667 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline