Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840681_m1
          See other MFAP4 GE Assays ›
          Gene Symbol:
          Mfap4
          Gene Name:
          microfibrillar-associated protein 4
          Gene Aliases:
          1110007F23Rik
          Chromosome Location:
          Chr.11: 61485378 - 61488706 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840681_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mfap4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029568.2 NP_083844.1 3 - 4 279 68
          GenBank mRNA AK003537.1 - 3 - 4 279 68
          AK075659.1 - 3 - 4 340 68
          BC022666.1 - 3 - 4 271 4224106 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000780.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mfap4

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s)

          cell adhesion
          UV protection
          regulation of collagen metabolic process
          extracellular fibril organization
          elastic fiber assembly
          cellular response to UV-B

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005470 AK075659.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005470 BC022666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005470 AK003537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005470 NM_029568.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718532 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131750 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005470 AK075659.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005470 BC022666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005470 AK003537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005470 NM_029568.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P195244 AK075659.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P195244 BC022666.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P195244 AK003537.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P195244 NM_029568.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27848628 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131750 - -
          Whole Mouse Genome G4122A A_51_P195244 AK075659.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195244 BC022666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195244 AK003537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195244 NM_029568.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106264 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131467 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131750 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09156507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09156491 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09156509 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09156494 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09156497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09156515 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09156511 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09156493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27848569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09156515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09480486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09156493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09554668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09308264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09462639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09462671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27848619 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376779 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376782 - -
          Affymetrix HT MG-430 PM Array Plate 1424010_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10376778 - -
          Mouse Genome 430 2.0 Array 1424010_at - -
          Mouse Genome 430 2.0 Array 1424010_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76293_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76293_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100006247.mm.1 NM_029568.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006247.mm.1 NM_029568.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006247.mm.1 TC1100000780.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006248.mm.1 NM_029568.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006248.mm.1 NM_029568.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006248.mm.1 TC1100000780.mm.1 MTA TC
          Mouse Expression Set 430 1424010_at - -
          Mouse Expression Set 430 1424010_PM_at - -
          Mouse Genome 430A 2.0 Array 1424010_at - -
          Mouse Genome 430A 2.0 Array 1424010_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424010_at - -
          Affymetrix HT Mouse Genome 430A Array 1424010_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 777324 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 520181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520181 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2633350 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2633350 - -
          mouse-6 v1.1 (Illumina) scl41467.7_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 520181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41467.7_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520181 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2633350 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2633350 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline