Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00847410_sH
          See other OLFR874 GE Assays ›
          Gene Symbol:
          Olfr874
          Gene Name:
          olfactory receptor 874
          Gene Aliases:
          MOR161-2
          Chromosome Location:
          Chr.9: 37743134 - 37747106 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00847410_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr874
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146882.2 NP_667093.2 1 38 135

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr874

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069632 NM_146882.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2120902 NM_146882.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2120902 NM_146882.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069632 NM_146882.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P122116 NM_146882.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432334 - -
          Whole Mouse Genome G4122A A_51_P122116 NM_146882.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107033 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105439 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07562848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07562844 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07929655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07464569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08099664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07511900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08099665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432319 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584353 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584352 - -
          Affymetrix Mouse Clariom S Array PSR0900002754.mm.1 NM_146882.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002754.mm.1 NM_146882.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002754.mm.1 TC0900000373.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 724539 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252965 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252965 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252965 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline