Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00848119_s1
          See other TCEB2 GE Assays ›
          Gene Symbol:
          Tceb2
          Gene Name:
          transcription elongation factor B (SIII), polypeptide 2
          Gene Aliases:
          0610040H15Rik
          Chromosome Location:
          Chr.17: 23824740 - 23829109 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00848119_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tceb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026305.2 NP_080581.1 4 433 84
          GenBank mRNA AK002868.1 - 4 434 84
          AK003277.1 - 4 425 84
          AK008534.2 - 4 436 84
          AK012254.1 - 4 426 84
          AK152950.1 - 5 504 84
          BC056983.1 - 3 418 6823854 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tceb2

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription elongation from RNA polymerase II promoter
          positive regulation of transcription from RNA polymerase II promoter
          transcription coactivator activity
          protein binding
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093905 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093906 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980932 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138780 AK003277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138780 AK008534.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138780 AK152950.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138780 BC056983.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138780 NM_026305.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458571 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42974 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136066 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138780 AK003277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138780 AK008534.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138780 AK152950.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138780 BC056983.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138780 NM_026305.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015692 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980932 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P397823 AK012254.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P397823 AK019181.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P397823 AK003277.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P397823 AK019218.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P397823 AK008477.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1182012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093951 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136066 - -
          Whole Mouse Genome G4122A A_51_P397823 AK012254.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397823 AK019181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397823 AK003277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397823 AK019218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397823 AK008477.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1182012 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138780 AK003277.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138780 AK008534.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138780 AK152950.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138780 BC056983.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138780 NM_026305.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136066 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13195120 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13195118 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13138918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13195119 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093906 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31093905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13222782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13463824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13195120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13121783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13156753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13463837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13222787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13491099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13222786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13121786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13121781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093882 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448382 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448381 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452891 - -
          Affymetrix HT MG-430 PM Array Plate 1428263_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10452890 - -
          Affymetrix Mouse Gene 1.1 ST Array 10448380 - -
          Mouse Genome 430 2.0 Array 1428263_a_at - -
          Mouse Genome 430 2.0 Array 1436949_PM_a_at - -
          Mouse Genome 430 2.0 Array 1436949_a_at - -
          Mouse Genome 430 2.0 Array 1428263_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR1700014070.mm.1 NM_026305.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014070.mm.1 NM_026305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014070.mm.1 TC1700001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014069.mm.1 NM_026305.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014069.mm.1 NM_026305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014069.mm.1 TC1700001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014072.mm.1 NM_026305.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014072.mm.1 NM_026305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014072.mm.1 TC1700001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014073.mm.1 NM_026305.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014073.mm.1 NM_026305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014073.mm.1 TC1700001606.mm.1 MTA TC
          Mouse Expression Set 430 1428263_PM_a_at - -
          Mouse Expression Set 430 1428263_a_at - -
          Mouse Expression Set 430 1436949_PM_a_at - -
          Mouse Expression Set 430 1436949_a_at - -
          Mouse Genome 430A 2.0 Array 1436949_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1428263_a_at - -
          Mouse Genome 430A 2.0 Array 1428263_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1436949_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428263_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428263_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 933131 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 840204 - -
          MouseWG-6 v2 Expression BeadChip 1340400 - -
          Illumina mouse-6 v1.0 expression beadchip 1340400 - -
          Illumina mouse-6 v1.0 expression beadchip 1450082 - -
          Illumina mouse-6 v1.0 expression beadchip 840204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340400 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744326 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658574 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2687706 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2687706 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744326 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2658574 - -
          mouse-6 v1.1 (Illumina) scl0067673.1_322-S - -
          mouse-6 v1.1 (Illumina) scl067673.2_0-S - -
          mouse-6 v1.1 (Illumina) scl50229.6.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450082 - -
          Illumina Sentrix mouse-6.v1 bead array 1340400 - -
          Illumina Sentrix mouse-6.v1 bead array 840204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067673.1_322-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50229.6.1_24-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067673.2_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340400 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840204 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450082 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216504 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2744326 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2687706 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2687706 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216504 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2744326 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067673.2_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50229.6.1_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0067673.1_322-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline