Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00848465_s1
          See other OLFR512 GE Assays ›
          Gene Symbol:
          Olfr512
          Gene Name:
          olfactory receptor 512
          Gene Aliases:
          MOR268-3
          Chromosome Location:
          Chr.7: 108713355 - 108714335 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00848465_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr512
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146724.1 NP_666935.1 1 280 135
          GenBank mRNA BC116968.1 - 1 315 40126287 135

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr512

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161542 NM_146724.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161542 BC116968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161542 NM_146724.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161542 BC116968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161542 NM_146724.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161542 BC116968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161542 NM_146724.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161542 BC116968.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P161542 NM_146724.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P161542 BC116968.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P161542 NM_146724.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P161542 BC116968.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404290 - -
          Whole Mouse Genome G4122A A_51_P161542 NM_146724.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161542 BC116968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161542 NM_146724.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161542 BC116968.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130470 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06474582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06652528 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06652527 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06652526 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06652536 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06652541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06556863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06474576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06540932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06094778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06556855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404326 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556163 - -
          Affymetrix Mouse Gene 1.0 ST Array 10556164 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556162 - -
          Affymetrix Mouse Clariom S Array PSR0700013289.mm.1 TC0700001604.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013290.mm.1 TC0700001604.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013290.mm.1 NM_146724.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013290.mm.1 NM_146724.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013291.mm.1 TC0700001604.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 691587 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2640348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002640348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230268 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230268 - -
          mouse-6 v1.1 (Illumina) scl0258719.1_122-S - -
          Illumina Sentrix mouse-6.v1 bead array 2640348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258719.1_122-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2640348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2902804 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230268 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230268 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2902804 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline