Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00850344_g1
          See other TOMM22 GE Assays ›
          Gene Symbol:
          Tomm22
          Gene Name:
          translocase of outer mitochondrial membrane 22 homolog (yeast)
          Gene Aliases:
          2310047D01, Tom22
          Chromosome Location:
          Chr.15: 79670868 - 79672862 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00850344_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tomm22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172609.3 NP_766197.2 4 548 150
          GenBank mRNA AK008133.1 - 4 551 150
          AK009868.1 - 4 548 150
          AK013471.1 - 4 537 150
          AK049442.1 - 536 150
          AK150360.1 - 4 536 150
          AK150787.1 - 4 545 150
          AK152843.1 - 4 536 150
          AK167803.1 - 4 536 150
          BC056920.1 - 4 530 6819748 150

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tomm22

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          protein transport
          protein import into mitochondrial outer membrane
          protein binding
          protein transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30367791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30367786 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30367790 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30367785 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968276 BC056920.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968276 AK049442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968276 AK150360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968276 AK150787.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968276 AK152843.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 820414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159687 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143735 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968276 BC056920.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968276 AK049442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968276 AK150360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968276 AK150787.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968276 AK152843.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P105339 BC056920.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P105339 AK049442.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P105339 AK150360.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P105339 AK150787.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P105339 AK152843.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30367797 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143735 - -
          Whole Mouse Genome G4122A A_51_P105339 BC056920.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105339 AK049442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105339 AK150360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105339 AK150787.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105339 AK152843.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100097 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143735 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12349233 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30367791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30367786 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30367785 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30367790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30367785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11968488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12102979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11929586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12218593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11929592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12307683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30367802 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514925 - -
          Affymetrix HT MG-430 PM Array Plate 1434420_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1429103_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10514924 - -
          Affymetrix Mouse Gene 1.1 ST Array 10425299 - -
          Mouse Genome 430 2.0 Array 1434420_PM_x_at - -
          Mouse Genome 430 2.0 Array 1429103_at - -
          Mouse Genome 430 2.0 Array 1434420_x_at - -
          Mouse Genome 430 2.0 Array 1429103_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223696_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223696_at - -
          Affymetrix Mouse Clariom S Array PSR1500005085.mm.1 NM_172609.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005085.mm.1 NM_172609.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005085.mm.1 TC1500000678.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005086.mm.1 NM_172609.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005086.mm.1 NM_172609.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005086.mm.1 TC1500000678.mm.1 MTA TC
          Mouse Expression Set 430 1434420_x_at - -
          Mouse Expression Set 430 1429103_at - -
          Mouse Expression Set 430 1429103_PM_at - -
          Mouse Expression Set 430 1434420_PM_x_at - -
          Murine Genome U74v2 Set 171094_i_at - -
          Murine Genome U74v2 Set 171094_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1434420_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1434420_x_at - -
          Mouse Genome 430A 2.0 Array 1429103_at - -
          Mouse Genome 430A 2.0 Array 1429103_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434420_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1434420_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1429103_at - -
          Affymetrix HT Mouse Genome 430A Array 1429103_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 331749 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104850341 - -
          Illumina mouse-6 v1.0 expression beadchip 104850341 - -
          Illumina mouse-6 v1.0 expression beadchip 1240142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001240142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2507182 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2444077 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2507182 - -
          mouse-6 v1.1 (Illumina) scl00223696.2_201-S - -
          mouse-6 v1.1 (Illumina) scl47742.5_273-S - -
          Illumina Sentrix mouse-6.v1 bead array 104850341 - -
          Illumina Sentrix mouse-6.v1 bead array 1240142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00223696.2_201-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104850341 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2507182 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2507182 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2444077 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00223696.2_201-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline