Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00851543_g1
          See other VMN1R27 GE Assays ›
          Gene Symbol:
          Vmn1r27
          Gene Name:
          vomeronasal 1 receptor 27
          Gene Aliases:
          V1rc33
          Chromosome Location:
          Chr.6: 58215106 - 58216017 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00851543_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134436.2 NP_602318.2 1 831 133

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r27

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227594 NM_134436.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227594 NM_134436.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P227594 NM_134436.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P227594 NM_134436.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P227594 NM_134436.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P227594 NM_134436.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24481457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24481443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24481483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24481442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24481480 - -
          Whole Mouse Genome G4122A A_51_P227594 NM_134436.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227594 NM_134436.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24481426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24481443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24481442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05668224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05400085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05760784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24481480 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538589 - -
          Affymetrix Mouse Gene 1.1 ST Array 10545036 - -
          Affymetrix Mouse Gene 1.1 ST Array 10538588 - -
          Affymetrix Mouse Clariom S Array PSR0600018398.mm.1 TC0600002369.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018398.mm.1 NM_134436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018398.mm.1 NM_134436.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 551320 - -
          ABI Mouse Genome Survey Microarray 724199 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1050537 - -
          Illumina mouse-6 v1.0 expression beadchip 102640333 - -
          Illumina mouse-6 v1.0 expression beadchip 1050537 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001050537 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257730 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257730 - -
          mouse-6 v1.1 (Illumina) scl0171206.1_225-S - -
          mouse-6 v1.1 (Illumina) GI_21717792-S - -
          Illumina Sentrix mouse-6.v1 bead array 102640333 - -
          Illumina Sentrix mouse-6.v1 bead array 1050537 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0171206.1_225-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102640333 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1050537 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257730 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257730 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline