Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01144495_m1
          See other RWDD2A GE Assays ›
          Gene Symbol:
          Rwdd2a
          Gene Name:
          RWD domain containing 2A
          Gene Aliases:
          1700030C20Rik, AI848608, Rwdd2
          Chromosome Location:
          Chr.9: 86571988 - 86574899 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01144495_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rwdd2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145968.1 NP_001139440.1 2 - 3 263 70
          NM_001145969.1 NP_001139441.1 2 - 3 297 70
          NM_027100.2 NP_081376.2 2 - 3 392 70
          GenBank mRNA AK006533.1 - 2 - 3 391 70
          BC115691.1 - 2 - 3 392 40104313 70
          BC115692.1 - 2 - 3 263 40104314 70
          CN836837.1 - 2 - 3 300 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001115.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rwdd2a

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26700014 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26700015 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 AK006533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 BC115691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 BC115692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 NM_001145969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 NM_027100.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 AK006533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 BC115691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 BC115692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 NM_001145969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 NM_027100.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563284 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165096 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 AK006533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 BC115691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 BC115692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 NM_001145969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 NM_027100.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 AK006533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 BC115691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 BC115692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 NM_001145969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 NM_027100.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700036 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165096 - -
          Whole Mouse Genome G4122A A_52_P554436 AK006533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 BC115691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 BC115692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 NM_001145969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 NM_027100.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 AK006533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 BC115691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 BC115692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 NM_001145969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 NM_027100.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137626 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165096 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07779078 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07779074 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07779077 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07779072 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26700014 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26700015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07965323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07779087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07779078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07991571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07843403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07965310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07779077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700044 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10587606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587605 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587609 - -
          Affymetrix HT MG-430 PM Array Plate 1428963_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10587604 - -
          Mouse Genome 430 2.0 Array 1428963_at - -
          Mouse Genome 430 2.0 Array 1428963_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69519_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69519_at - -
          Affymetrix Mouse Clariom S Array PSR0900008780.mm.1 NM_001145968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008780.mm.1 NM_001145968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008780.mm.1 NM_001145969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008780.mm.1 NM_001145969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008780.mm.1 NM_027100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008780.mm.1 NM_027100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008780.mm.1 TC0900001115.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008779.mm.1 NM_001145968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008779.mm.1 NM_001145968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008779.mm.1 NM_001145969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008779.mm.1 NM_001145969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008779.mm.1 NM_027100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008779.mm.1 NM_027100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008779.mm.1 TC0900001115.mm.1 MTA TC
          Mouse Expression Set 430 1428963_PM_at - -
          Mouse Expression Set 430 1428963_at - -
          Affymetrix HT Mouse Genome 430B Array 1428963_at - -
          Affymetrix HT Mouse Genome 430B Array 1428963_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 310349 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1660079 - -
          Illumina mouse-6 v1.0 expression beadchip 1660079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774528 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774528 - -
          mouse-6 v1.1 (Illumina) scl36646.3.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36646.3.1_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660079 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774528 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36646.3.1_24-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0