Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ankrd24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027480.3 | NP_081756.4 | 6 - 7 | 644 | 55 | |
GenBank mRNA | AK043638.1 | - | 5 - 6 | 1094 | 55 | |
AK173316.1 | - | 6 - 7 | 467 | 55 | ||
BC048236.1 | - | 5 - 6 | 549 | 6313999 | 55 | |
CB524981.1 | - | 3 - 4 | 173 | 55 | ||
CJ183294.1 | - | 3 - 4 | 359 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |