Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01156721_g1
          See other H19 GE Assays ›
          Gene Symbol:
          H19
          Gene Name:
          H19, imprinted maternally expressed transcript
          Gene Aliases:
          AI747191
          Chromosome Location:
          Chr.7: 142575532 - 142578146 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01156721_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK145379.1 - 3 - 4 1540 78
          CV561038.1 - 3 - 4 697 78

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961127 NR_001592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961127 AK145379.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961125 NR_001592.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 514405 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116993 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961127 NR_001592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961127 AK145379.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116993 - -
          Whole Mouse Genome G4122A A_51_P142196 NR_001592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P142196 AK145379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P571350 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103608 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25595797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25595794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25595799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25595800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25595793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25595796 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569343 - -
          Affymetrix HT MG-430 PM Array Plate 1448194_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569335 - -
          Affymetrix Mouse Gene 1.1 ST Array 10569341 - -
          Mouse Genome 430 2.0 Array 1448194_PM_a_at - -
          Mouse Genome 430 2.0 Array 1448194_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14955_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14955_at - -
          Affymetrix Murine Genome 6500A Array Msa14550_at - -
          Mouse Expression Set 430 1448194_a_at - -
          Mouse Expression Set 430 1448194_PM_a_at - -
          Murine Genome U74v2 Set 93028_PM_at - -
          Murine Genome U74v2 Set 93028_at - -
          Mouse Genome 430A 2.0 Array 1448194_a_at - -
          Mouse Genome 430A 2.0 Array 1448194_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448194_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448194_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106840152 - -
          Illumina mouse-6 v1.0 expression beadchip 106840152 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256343 - -
          mouse-6 v1.1 (Illumina) scl30470.4_40-S - -
          Illumina Sentrix mouse-6.v1 bead array 106840152 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106840152 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2906728 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2906728 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256343 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0