Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01159323_g1
          See other G6B GE Assays ›
          Gene Symbol:
          G6b
          Gene Name:
          immunoreceptor tyrosine-based inhibitory motif (ITIM) containing platelet receptor
          Gene Aliases:
          AU023871, C6orf25, Ng31
          Chromosome Location:
          Chr.17: 35062693 - 35066785 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01159323_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: G6b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033221.3 NP_001028393.1 1 - 2 76 92
          NM_001191012.1 NP_001177941.1 1 - 2 76 92
          GenBank mRNA AK137516.1 - 1 - 2 77 92
          GU269270.1 - 1 - 2 62 92

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          integrin-mediated signaling pathway
          blood coagulation
          platelet formation
          megakaryocyte development
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157365 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157369 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157368 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157367 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157366 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986531 NM_001033221.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986531 NM_001191012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986531 AK137516.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63829 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117723 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986531 NM_001033221.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986531 NM_001191012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986531 AK137516.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157342 - -
          Whole Mouse Genome G4122A A_52_P205473 GU269270.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P205473 NM_001033221.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P205473 NM_001191012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P205473 AK137516.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568648 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13249326 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568642 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568646 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157366 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157367 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157368 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157365 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13249323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13249325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13249326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157374 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450416 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450415 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450412 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161372 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161372 - -
          mouse-6 v1.1 (Illumina) GI_85701725-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161372 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161372 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline