Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164808_m1
          See other 4930599N23RIK GE Assays ›
          Gene Symbol:
          4930599N23Rik
          Gene Name:
          RIKEN cDNA 4930599N23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 39118473 - 39148312 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164808_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930599N23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045813.1 2 - 3 463 66
          GenBank mRNA AK016417.1 - 2 - 3 463 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000390.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100586 AK042039.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106267 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168560 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123312 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100586 AK042039.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05870652 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123312 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05870639 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168560 - -
          Whole Mouse Genome G4122A A_52_P359071 AK016417.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P359071 AK042039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241640 AK016417.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123312 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168560 - -
          Array Name Probe IDs Targets Target Type
          Mouse Genome 430 2.0 Array 1456150_PM_at - -
          Mouse Genome 430 2.0 Array 1456150_at - -
          Affymetrix Mouse Clariom S Array PSR0600003021.mm.1 TC0600000390.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003021.mm.1 NR_045813.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003021.mm.1 NR_045813.1 Refseq
          Mouse Expression Set 430 1456150_at - -
          Mouse Expression Set 430 1456150_PM_at - -
          Murine Genome U74v2 Set 136543_PM_i_at - -
          Murine Genome U74v2 Set 136543_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 721067 - -
          ABI Mouse Genome Survey Microarray 800239 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 104610403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257546 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258612 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257546 - -
          Illumina Sentrix mouse-6.v1 bead array 104610403 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104610403 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257546 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258612 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0