Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01165516_s1
          See other HIST1H4I GE Assays ›
          Gene Symbol:
          Hist1h4i
          Gene Name:
          histone cluster 1, H4i
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 22040960 - 22041362 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01165516_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h4i
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175656.3 NP_783587.1 1 351 71
          GenBank mRNA AK011560.1 - 1 352 71
          BC019757.1 - 1 341 71
          BC052219.1 - 1 324 71
          BC092144.1 - 1 341 30612926 71
          BC125598.1 - 1 335 40130113 71
          BC125600.1 - 1 335 40130115 71

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hist1h4i

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          DNA replication-dependent nucleosome assembly
          DNA replication-independent nucleosome assembly
          negative regulation of megakaryocyte differentiation
          protein heterotetramerization
          histone binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28876028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28876022 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505521 BC019757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505521 BC019757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174198 BC092144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174198 NM_175656.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158697 BC092144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158697 BC125600.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158697 NM_175656.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158697 AK011560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158697 BC052219.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163195 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158697 BC092144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158697 BC125600.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158697 NM_175656.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158697 AK011560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158697 BC052219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174198 BC092144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174198 NM_175656.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28876034 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163195 - -
          Whole Mouse Genome G4122A A_51_P505521 BC019757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505521 BC019757.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116654 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10997971 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11087603 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11087618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11087596 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10997968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28876028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28876022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28875962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28876066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11000741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10456847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11121524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28876034 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10408093 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403958 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403957 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408113 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408092 - -
          Mouse Genome 430 2.0 Array 1424854_at - -
          Mouse Genome 430 2.0 Array 1424854_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319158_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319158_at - -
          Affymetrix Mouse Clariom S Array PSR1300009921.mm.1 NM_175656.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009921.mm.1 TC1300001575.mm.1 MTA TC
          Mouse Expression Set 430 1424854_PM_at - -
          Mouse Expression Set 430 1422948_PM_s_at - -
          Mouse Expression Set 430 1422947_at - -
          Mouse Expression Set 430 1424854_at - -
          Mouse Expression Set 430 1428014_PM_at - -
          Mouse Expression Set 430 1428014_at - -
          Mouse Expression Set 430 1422947_PM_at - -
          Mouse Expression Set 430 1422948_s_at - -
          Mouse Expression Set 430 1431615_at - -
          Mouse Expression Set 430 1431658_at - -
          Mouse Expression Set 430 1431658_PM_at - -
          Mouse Expression Set 430 1431615_PM_at - -
          Mouse Genome 430A 2.0 Array 1428014_at - -
          Mouse Genome 430A 2.0 Array 1422948_s_at - -
          Mouse Genome 430A 2.0 Array 1422948_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1424854_PM_at - -
          Mouse Genome 430A 2.0 Array 1422947_PM_at - -
          Mouse Genome 430A 2.0 Array 1428014_PM_at - -
          Mouse Genome 430A 2.0 Array 1422947_at - -
          Mouse Genome 430A 2.0 Array 1424854_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 681625 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2190450 - -
          Illumina mouse-6 v1.0 expression beadchip 2190450 - -
          Illumina mouse-6 v1.0 expression beadchip 106550022 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002190450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232524 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240763 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246576 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232524 - -
          mouse-6 v1.1 (Illumina) scl44225.1.12_219-S - -
          mouse-6 v1.1 (Illumina) scl00319158.2_61-S - -
          Illumina Sentrix mouse-6.v1 bead array 2190450 - -
          Illumina Sentrix mouse-6.v1 bead array 106550022 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00319158.2_61-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190450 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106550022 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232524 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232524 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline