Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01172887_m1
          See other CACTIN GE Assays ›
          Gene Symbol:
          Cactin
          Gene Name:
          cactin, spliceosome C complex subunit
          Gene Aliases:
          2510012J08Rik
          Chromosome Location:
          Chr.10: 81321103 - 81326251 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01172887_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cactin
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027381.2 NP_081657.1 1 - 2 345 87
          GenBank mRNA AY934504.1 - 1 - 2 251 87
          BC150986.1 - 1 - 2 345 9087899 87
          BC151180.1 - 1 - 2 345 9088069 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000893.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          immune system process
          mRNA processing
          multicellular organism development
          RNA splicing
          negative regulation of lipopolysaccharide-mediated signaling pathway
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of interferon-beta production
          negative regulation of interleukin-8 production
          negative regulation of tumor necrosis factor production
          negative regulation of toll-like receptor signaling pathway
          innate immune response
          negative regulation of type I interferon-mediated signaling pathway
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288888 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288887 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288893 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288889 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288855 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288890 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288853 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288851 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288850 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288894 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288892 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288864 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288856 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288886 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288891 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281806 BY651766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281806 AY934504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281806 BC023696.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281806 AK010963.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281806 NM_027381.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281806 BY651766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281806 AY934504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281806 BC023696.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281806 AK010963.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281806 NM_027381.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 588883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728956 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139320 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131674 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281806 BY651766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281806 AY934504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281806 BC023696.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281806 AK010963.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281806 NM_027381.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281806 BY651766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281806 AY934504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281806 BC023696.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281806 AK010963.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281806 NM_027381.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131674 - -
          Whole Mouse Genome G4122A A_52_P482849 BC151180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482849 AY934504.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482849 BC023696.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482849 BC150986.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482849 AK010963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281806 BY651766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281806 AY934504.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281806 BC023696.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281806 AK010963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281806 NM_027381.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281806 BY651766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281806 AY934504.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281806 BC023696.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281806 AK010963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281806 NM_027381.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131674 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288890 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288893 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288892 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288856 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288851 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288853 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288894 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288864 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288850 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288888 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288891 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288886 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288889 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288855 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08397144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08294539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08694554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08397138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08893479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08694558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08397161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08397158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08949982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08893478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288872 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10365091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365093 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365097 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365094 - -
          Affymetrix HT MG-430 PM Array Plate 1452760_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10365088 - -
          Mouse Genome 430 2.0 Array 1452760_at - -
          Mouse Genome 430 2.0 Array 1452760_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70312_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70312_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000007034.mm.1 NM_027381.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007034.mm.1 NM_027381.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007034.mm.1 TC1000000893.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007035.mm.1 NM_027381.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007035.mm.1 NM_027381.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007035.mm.1 TC1000000893.mm.1 MTA TC
          Mouse Expression Set 430 1452760_PM_at - -
          Mouse Expression Set 430 1452760_at - -
          Murine Genome U74v2 Set 167815_f_at - -
          Murine Genome U74v2 Set 128770_PM_f_at - -
          Murine Genome U74v2 Set 128770_f_at - -
          Murine Genome U74v2 Set 167815_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1452760_at - -
          Affymetrix HT Mouse Genome 430B Array 1452760_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 838398 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255323 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2429710 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2974280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255323 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2974280 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline