Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175430_m1
          See other SEPT5 GE Assays ›
          Gene Symbol:
          Sept5
          Gene Name:
          septin 5
          Gene Aliases:
          Cdcrel-1, Cdcrel1, Pnutl1
          Chromosome Location:
          Chr.16: 18621811 - 18629938 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175430_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sept5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_213614.2 NP_998779.2 8 - 9 804 109
          GenBank mRNA AF033350.1 - 6 - 7 651 109
          AK134700.1 - 8 - 9 741 109
          BC059848.1 - 6 - 7 682 109
          BC079631.1 - 2 - 3 118 109
          BC141073.1 - 8 - 9 814 9055685 109
          BC145331.1 - 8 - 9 814 9053887 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sept5

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          regulation of exocytosis
          positive regulation of exocytosis
          cell division
          nucleotide binding
          protein binding
          GTP binding
          GDP binding
          syntaxin binding
          phosphatidylinositol binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039881 BC094347.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039881 BC141073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039881 BC145331.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039881 BC145332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039881 BC059848.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206384 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63577 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039881 BC094347.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039881 BC141073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039881 BC145331.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039881 BC145332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039881 BC059848.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P464530 BC094347.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P464530 BC059848.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P464530 AK134700.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P464530 BC079631.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P464530 NM_213614.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P21353 BC094347.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P21353 BC141073.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P21353 BC145331.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P21353 BC145332.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P21353 BC059848.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464530 BC094347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464530 BC059848.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464530 AK134700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464530 BC079631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464530 NM_213614.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21353 BC094347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21353 BC141073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21353 BC145331.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21353 BC145332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21353 BC059848.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12637531 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12637553 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12637542 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12637534 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12637536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438369 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438368 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438372 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438370 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438367 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438373 - -
          Affymetrix Mouse Gene 1.1 ST Array 10438358 - -
          Mouse Genome 430 2.0 Array 1452357_at - -
          Mouse Genome 430 2.0 Array 1452357_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600010883.mm.1 NM_213614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010883.mm.1 NM_213614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010883.mm.1 TC1600001319.mm.1 MTA TC
          Mouse Expression Set 430 1452357_PM_at - -
          Mouse Expression Set 430 1452357_at - -
          Murine Genome U74v2 Set 96337_at - -
          Murine Genome U74v2 Set 96337_PM_at - -
          Mouse Genome 430A 2.0 Array 1452357_at - -
          Mouse Genome 430A 2.0 Array 1452357_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 836508 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 460725 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2828265 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2828265 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline