Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01177636_m1
          See other TMPRSS9 GE Assays ›
          Gene Symbol:
          Tmprss9
          Gene Name:
          transmembrane protease, serine 9
          Gene Aliases:
          Serase-1B
          Chromosome Location:
          Chr.10: 80871117 - 80899495 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01177636_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmprss9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081688.2 NP_001075157.2 12 - 13 2022 79
          GenBank mRNA BC158102.1 - 12 - 13 2118 9007387 79
          BC172169.1 - 12 - 13 2118 9054839 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000875.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          plasminogen activation
          serine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976949 AB109391.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976953 NM_001081688.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530696 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608342 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 758641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 100687 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976953 NM_001081688.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08936394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08707478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08276998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08767629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08672269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08887315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08672264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08433696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08767623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08340631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08354259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08339748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08392838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08767622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08690046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08773651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08670392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08670391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08707481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08392836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08773653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08670394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08660698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08783259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08672262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08254734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08887318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08476904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08527856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08628876 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364945 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364942 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364934 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364940 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364939 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364941 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364946 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364937 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364943 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364936 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364935 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364938 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364944 - -
          Affymetrix Mouse Gene 1.1 ST Array 10364932 - -
          Affymetrix Mouse Clariom S Array PSR1000006753.mm.1 NM_001081688.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006753.mm.1 NM_001081688.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006753.mm.1 TC1000000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000006752.mm.1 NM_001081688.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006752.mm.1 NM_001081688.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006752.mm.1 TC1000000875.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 361317 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_3142104 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3142104 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline