Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179943_m1
          See other TMEM50A GE Assays ›
          Gene Symbol:
          Tmem50a
          Gene Name:
          transmembrane protein 50A
          Gene Aliases:
          3200001F09Rik, CAM, Smp1
          Chromosome Location:
          Chr.4: 134897849 - 134915030 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179943_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem50a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027935.2 NP_082211.1 6 - 7 475 84
          GenBank mRNA AK014282.1 - 6 - 7 545 84
          AK147069.1 - 6 - 7 511 84
          AK152630.1 - 6 - 7 559 84
          AK152654.1 - 6 - 7 530 84
          AK157214.1 - 6 - 7 548 84
          AK164989.1 - 6 - 7 541 84
          AK165636.1 - 6 - 7 524 84
          BC094256.1 - 6 - 7 475 6820666 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003749.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem50a

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325517 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325520 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325519 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325518 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325558 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058497 AK164989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058497 AK165636.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058497 BC094256.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058497 NM_027935.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058497 AK014282.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391927 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824865 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506610 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396844 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160413 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102726 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113695 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058497 AK164989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058497 AK165636.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058497 BC094256.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058497 NM_027935.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058497 AK014282.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325509 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113695 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102726 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05413141 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05413150 - -
          Whole Mouse Genome G4122A A_51_P414160 AK164989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414160 AK165636.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414160 BC094256.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414160 NM_027935.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414160 AK014282.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124377 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113695 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102726 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04017320 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03376770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03778617 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03785945 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03376755 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04017319 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03607910 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325517 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325558 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325518 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325519 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03721024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03960028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03728204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03376755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03376759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03376766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03960025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03376769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03721023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03871307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03210725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03721019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03871297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03869238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03376756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03871290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03721027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03651770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03498219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03498220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03547707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325507 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10517333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517332 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517331 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517330 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517335 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517329 - -
          Affymetrix HT MG-430 PM Array Plate 1453155_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10517328 - -
          Mouse Genome 430 2.0 Array 1453155_PM_at - -
          Mouse Genome 430 2.0 Array 1453155_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71817_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71817_at - -
          Affymetrix Mouse Clariom S Array PSR0400030886.mm.1 TC0400003749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030886.mm.1 NM_027935.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030886.mm.1 NM_027935.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030887.mm.1 TC0400003749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030887.mm.1 NM_027935.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030887.mm.1 NM_027935.2 Refseq
          Mouse Expression Set 430 1453155_PM_at - -
          Mouse Expression Set 430 1453155_at - -
          Affymetrix HT Mouse Genome 430B Array 1453155_at - -
          Affymetrix HT Mouse Genome 430B Array 1453155_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 498407 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2690440 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690440 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2627286 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2627286 - -
          mouse-6 v1.1 (Illumina) scl23678.5.1_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 2690440 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23678.5.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690440 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2819679 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2819679 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl23678.5.1_21-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline