| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600023761.mm.1 |
TC0600003067.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023761.mm.1 |
NM_027664.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023761.mm.1 |
NM_027664.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023770.mm.1 |
TC0600003067.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023770.mm.1 |
NM_027664.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023770.mm.1 |
NM_027664.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023762.mm.1 |
TC0600003067.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023762.mm.1 |
NM_027664.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023762.mm.1 |
NM_027664.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023763.mm.1 |
TC0600003067.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023763.mm.1 |
NM_027664.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023763.mm.1 |
NM_027664.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023764.mm.1 |
TC0600003067.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023764.mm.1 |
NM_027664.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023764.mm.1 |
NM_027664.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023768.mm.1 |
TC0600003067.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023768.mm.1 |
NM_027664.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023768.mm.1 |
NM_027664.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023769.mm.1 |
TC0600003067.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023769.mm.1 |
NM_027664.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023769.mm.1 |
NM_027664.1 |
Refseq |