Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01188251_m1
          See other RSPO3 GE Assays ›
          Gene Symbol:
          Rspo3
          Gene Name:
          R-spondin 3 homolog (Xenopus laevis)
          Gene Aliases:
          2810459H04Rik, AW742308, Cristin1, Thsd2
          Chromosome Location:
          Chr.10: 29453106 - 29535867 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01188251_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rspo3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028351.3 NP_082627.3 2 - 3 828 140
          GenBank mRNA AB055811.1 - 2 - 3 786 140
          AK013366.1 - 2 - 3 777 140
          AK076308.1 - 775 140
          AK133916.1 - 2 - 3 829 140
          AY864332.1 - 2 - 3 289 140
          BC103794.1 - 2 - 3 762 6517974 140
          BC145929.1 - 2 - 3 433 40131232 140
          BC145931.1 - 2 - 3 433 40131234 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001936.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rspo3

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          Wnt signaling pathway
          response to stimulus
          branching involved in labyrinthine layer morphogenesis
          positive regulation of canonical Wnt signaling pathway
          receptor binding
          frizzled binding
          heparin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000013946.mm.1 NM_028351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013946.mm.1 NM_028351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013946.mm.1 TC1000001936.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013944.mm.1 NM_028351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013944.mm.1 NM_028351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013944.mm.1 TC1000001936.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013947.mm.1 NM_028351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013947.mm.1 NM_028351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013947.mm.1 TC1000001936.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013948.mm.1 NM_028351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013948.mm.1 NM_028351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013948.mm.1 TC1000001936.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013945.mm.1 NM_028351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013945.mm.1 NM_028351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013945.mm.1 TC1000001936.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 448130 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline