Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01204930_m1
          See other 2300005B03RIK GE Assays ›
          Gene Symbol:
          2300005B03Rik
          Gene Name:
          RIKEN cDNA 2300005B03 gene
          Gene Aliases:
          SLURP-2, Slurp2
          Chromosome Location:
          Chr.15: 74742839 - 74746687 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01204930_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2300005B03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081961.1 NP_001075430.1 1 - 2 60 64
          GenBank mRNA AB272582.1 - 1 - 2 48 64
          BC115610.1 - 1 - 2 60 40102714 64
          BC115611.1 - 1 - 2 60 40102716 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001716.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165234 BC115610.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165234 BC115611.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165234 NM_001081961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165234 AK009031.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 702606 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2165234 BC115610.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2165234 BC115611.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2165234 NM_001081961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2165234 AK009031.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12199973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12199980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12136198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12136203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12136209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339357 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10429512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429513 - -
          Affymetrix Mouse Gene 1.1 ST Array 10429510 - -
          Affymetrix Mouse Clariom S Array PSR1500012962.mm.1 NM_001081961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012962.mm.1 NM_001081961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012962.mm.1 TC1500001716.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012964.mm.1 NM_001081961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012964.mm.1 NM_001081961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012964.mm.1 TC1500001716.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3840047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2700364 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2700364 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2700364 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline