Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vsig10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033311.3 | NP_001028483.2 | 2 - 3 | 316 | 70 | |
GenBank mRNA | AK138152.1 | - | 2 - 3 | 265 | 70 | |
CJ057085.1 | - | 2 - 3 | 265 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Inventoried Probe spans exons |