Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01212294_m1
          See other RASL11A GE Assays ›
          Gene Symbol:
          Rasl11a
          Gene Name:
          RAS-like, family 11, member A
          Gene Aliases:
          1110065D03Rik, AI852031
          Chromosome Location:
          Chr.5: 146845071 - 146847726 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01212294_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rasl11a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026864.1 NP_081140.1 3 - 4 403 88
          GenBank mRNA AK004371.1 - 3 - 4 357 88
          BC118009.1 - 1518 40102480 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001796.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          signal transduction
          small GTPase mediated signal transduction
          positive regulation of transcription from RNA polymerase I promoter
          nucleotide binding
          protein binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170389 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170393 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170391 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170392 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170390 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170387 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P340699 NM_026864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P340699 AK004371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P340699 BC118009.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P340699 NM_026864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P340699 AK004371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P340699 BC118009.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672457 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P340699 NM_026864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P340699 AK004371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P340699 BC118009.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P340699 NM_026864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P340699 AK004371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P340699 BC118009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340699 NM_026864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340699 AK004371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340699 BC118009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340699 NM_026864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340699 AK004371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340699 BC118009.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129480 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170392 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170389 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170391 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170393 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170387 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24170390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24170355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04479677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04598603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04250245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04764440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04598601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04931905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04612610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04164296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04876872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04612609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04616193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04450479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04899050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04293292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04367120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04272216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04132838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04103394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04367122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04612607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04293295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04164300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04293291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04616191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04764441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04350791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04132841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04925857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04303880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04479676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04598600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24170356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04497849 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10527539 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527542 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527545 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527543 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527541 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527540 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527544 - -
          Affymetrix HT MG-430 PM Array Plate 1429444_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10527538 - -
          Mouse Genome 430 2.0 Array 1429444_PM_at - -
          Mouse Genome 430 2.0 Array 1429444_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68895_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68895_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500016629.mm.1 TC0500001796.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016629.mm.1 NM_026864.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016629.mm.1 NM_026864.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016628.mm.1 TC0500001796.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016628.mm.1 NM_026864.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016628.mm.1 NM_026864.1 Refseq
          Mouse Expression Set 430 1429444_at - -
          Mouse Expression Set 430 1429444_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429444_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429444_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 575496 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2755936 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2932662 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2932662 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline