Search
Search

Taf4a| Gene Symbol: Taf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081092.1 | NP_001074561.1 | 8 - 9 | 2238 | 62 | |
| GenBank mRNA | AK136225.1 | - | 1 - 2 | 82 | 62 | |
| AY038601.1 | - | 8 - 9 | 1094 | 62 | ||
Set Membership: |
|