Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01212548_m1
          See other RASL11B GE Assays ›
          Gene Symbol:
          Rasl11b
          Gene Name:
          RAS-like, family 11, member B
          Gene Aliases:
          1190017B18Rik
          Chromosome Location:
          Chr.5: 74195326 - 74199477 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01212548_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rasl11b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026878.1 NP_081154.1 3 - 4 400 100
          GenBank mRNA BC008101.1 - 3 - 4 430 3156762 100
          BC083068.1 - 3 - 4 400 6308926 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000699.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          small GTPase mediated signal transduction
          nucleotide binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801768 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801770 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801773 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801767 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801771 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801769 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801766 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293938 BC008101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293938 BC083068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293938 AK004534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293938 NM_026878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293938 BC008101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293938 BC083068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293938 AK004534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293938 NM_026878.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925017 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311827 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142966 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293938 BC008101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293938 BC083068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293938 AK004534.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293938 NM_026878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293938 BC008101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293938 BC083068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293938 AK004534.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293938 NM_026878.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293938 BC008101.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293938 BC083068.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293938 AK004534.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293938 NM_026878.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293938 BC008101.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293938 BC083068.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293938 AK004534.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293938 NM_026878.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142966 - -
          Whole Mouse Genome G4122A A_51_P293938 BC008101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293938 BC083068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293938 AK004534.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293938 NM_026878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293938 BC008101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293938 BC083068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293938 AK004534.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293938 NM_026878.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120806 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142966 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04388945 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04388933 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04388922 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04388924 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04388934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04388947 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04388944 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801769 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801770 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801773 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801767 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801766 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801768 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23801771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23801779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04388934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04403878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04940782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04388926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04388944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04388935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04909390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04806638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04407531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04909397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04617921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04403867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04940787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04909396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23801719 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10522471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522469 - -
          Affymetrix HT MG-430 PM Array Plate 1423854_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10522467 - -
          Mouse Genome 430 2.0 Array 1423854_a_at - -
          Mouse Genome 430 2.0 Array 1423854_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68939_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68939_at - -
          Affymetrix Mouse Clariom S Array PSR0500006174.mm.1 TC0500000699.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500006174.mm.1 NM_026878.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500006174.mm.1 NM_026878.1 Refseq
          Mouse Expression Set 430 1423854_a_at - -
          Mouse Expression Set 430 1423854_PM_a_at - -
          Murine Genome U74v2 Set 95418_at - -
          Murine Genome U74v2 Set 95418_PM_at - -
          Mouse Genome 430A 2.0 Array 1423854_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1423854_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423854_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423854_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 773682 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101230279 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2537404 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2706819 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2706819 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2793062 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2793062 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline