Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01227681_m1
          See other CDC42BPG GE Assays ›
          Gene Symbol:
          Cdc42bpg
          Gene Name:
          CDC42 binding protein kinase gamma (DMPK-like)
          Gene Aliases:
          BC046418, MRCKgamma
          Chromosome Location:
          Chr.19: 6306457 - 6325655 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01227681_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdc42bpg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033342.1 NP_001028514.1 36 - 37 4859 66
          GenBank mRNA AK133191.1 - 13 - 14 1931 66
          BC046418.1 - 19 - 20 2507 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000112.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          phosphorylation
          intracellular signal transduction
          nucleotide binding
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P105765 AK133191.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P105765 BC046418.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P105765 NM_001033342.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P105765 AK133191.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P105765 BC046418.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P105765 NM_001033342.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471025 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28234 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571541 BC017159.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102192 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171428 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P105765 AK133191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P105765 BC046418.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P105765 NM_001033342.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P105765 AK133191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P105765 BC046418.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P105765 NM_001033342.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171428 - -
          Whole Mouse Genome G4122A A_52_P105765 AK133191.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P105765 BC046418.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P105765 NM_001033342.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P105765 AK133191.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P105765 BC046418.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P105765 NM_001033342.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14267207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14421575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14267210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14421565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14421567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14377097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14395914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14395924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14395918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14377109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14421631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14377087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14377092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14377080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14348789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14348791 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460860 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460870 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460862 - -
          Affymetrix HT MG-430 PM Array Plate 1436197_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460841 - -
          Mouse Genome 430 2.0 Array 1436197_PM_at - -
          Mouse Genome 430 2.0 Array 1436197_at - -
          Affymetrix Mouse Clariom S Array PSR1900001335.mm.1 NM_001033342.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001335.mm.1 NM_001033342.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001335.mm.1 TC1900000112.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900001336.mm.1 NM_001033342.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001336.mm.1 NM_001033342.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001336.mm.1 TC1900000112.mm.1 MTA TC
          Mouse Expression Set 430 1436197_PM_at - -
          Mouse Expression Set 430 1436197_at - -
          Affymetrix HT Mouse Genome 430B Array 1436197_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436197_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 534547 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2531530 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0