Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01246265_m1
          See other CYB5D2 GE Assays ›
          Gene Symbol:
          Cyb5d2
          Gene Name:
          cytochrome b5 domain containing 2
          Gene Aliases:
          9330151E16Rik, Gm2
          Chromosome Location:
          Chr.11: 72777232 - 72795839 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01246265_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyb5d2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024926.3 NP_001020097.2 2 - 3 469 80
          GenBank mRNA AK034043.1 - 2 - 3 167 80
          BC086682.1 - 2 - 3 435 30666422 80
          BC117721.1 - 1 - 2 51 80

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC117721.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyb5d2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          positive regulation of neuron differentiation
          heme binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913320 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913314 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913317 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913318 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913316 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913315 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913321 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956097 CD776759.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956097 CJ284318.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956097 NM_001024926.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956097 AK034043.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379581 AK003663.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656322 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496826 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 209698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 165464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60974 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103466 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956097 CD776759.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956097 CJ284318.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956097 NM_001024926.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956097 AK034043.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27913322 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02361208 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103466 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07394307 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103466 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09058885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09644564 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09058899 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09058895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913321 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913317 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913320 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913318 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913316 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913314 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27913315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27913240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09165407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09477532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09058899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09058895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09453780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27913299 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388228 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388230 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388233 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388231 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388232 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388229 - -
          Affymetrix HT MG-430 PM Array Plate 1436943_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1444065_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10388227 - -
          Mouse Genome 430 2.0 Array 1436943_PM_at - -
          Mouse Genome 430 2.0 Array 1444065_at - -
          Mouse Genome 430 2.0 Array 1444065_PM_at - -
          Mouse Genome 430 2.0 Array 1436943_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 192986_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 192986_at - -
          Affymetrix Mouse Clariom S Array PSR1100029233.mm.1 NM_001024926.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029233.mm.1 NM_001024926.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029233.mm.1 TC1100003152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029229.mm.1 NM_001024926.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029229.mm.1 NM_001024926.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029229.mm.1 TC1100003152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029228.mm.1 NM_001024926.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029228.mm.1 NM_001024926.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029228.mm.1 TC1100003152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029226.mm.1 NM_001024926.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029226.mm.1 NM_001024926.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029226.mm.1 TC1100003152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029227.mm.1 NM_001024926.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029227.mm.1 NM_001024926.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029227.mm.1 TC1100003152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029230.mm.1 NM_001024926.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029230.mm.1 NM_001024926.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029230.mm.1 TC1100003152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029232.mm.1 NM_001024926.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029232.mm.1 NM_001024926.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029232.mm.1 TC1100003152.mm.1 MTA TC
          Mouse Expression Set 430 1444065_at - -
          Mouse Expression Set 430 1436943_PM_at - -
          Mouse Expression Set 430 1444065_PM_at - -
          Mouse Expression Set 430 1436943_at - -
          Murine Genome U74v2 Set 171331_PM_at - -
          Murine Genome U74v2 Set 171331_at - -
          Affymetrix HT Mouse Genome 430B Array 1436943_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444065_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444065_at - -
          Affymetrix HT Mouse Genome 430B Array 1436943_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102360402 - -
          Illumina mouse-6 v1.0 expression beadchip 102360402 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238767 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218915 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2657818 - -
          Illumina Sentrix mouse-6.v1 bead array 102360402 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102360402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2957968 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2957968 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238767 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline