Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01250941_m1
          See other D10WSU102E GE Assays ›
          Gene Symbol:
          D10Wsu102e
          Gene Name:
          DNA segment, Chr 10, Wayne State University 102, expressed
          Gene Aliases:
          AI747614, C430041I18Rik
          Chromosome Location:
          Chr.10: 83360221 - 83368835 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01250941_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D10Wsu102e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026579.3 NP_080855.1 3 - 4 378 106
          GenBank mRNA AK020067.1 - 3 - 4 356 106
          AK049544.1 - 3 - 4 361 106
          AK128995.1 - 3 - 4 371 106
          AK147063.1 - 3 - 4 361 106
          AK160350.1 - 3 - 4 361 106
          AK162255.1 - 3 - 4 354 106
          BC035946.1 - 3 - 4 335 106
          BC050904.1 - 3 - 4 346 6416625 106
          BC094241.1 - 3 - 4 340 30610591 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000940.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene D10Wsu102e

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297060 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297058 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297057 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297053 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297054 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297052 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297055 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297056 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299739 BC050904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299739 BC094241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299739 BC035946.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299739 AK128995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299739 AK147063.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948951 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739445 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58638 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168414 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299739 BC050904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299739 BC094241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299739 BC035946.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299739 AK128995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299739 AK147063.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27297050 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168414 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07129022 - -
          Whole Mouse Genome G4122A A_52_P140663 BC050904.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P140663 BC094241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P140663 AK160350.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P140663 AK162255.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P140663 AK199731.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105565 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168414 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08621596 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08621597 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08621605 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08621590 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08621589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08621594 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297055 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297056 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297057 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297060 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297052 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297053 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297058 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27297054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27297131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08678336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08621586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08922409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08432732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08820039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08678327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08769636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08678337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08549262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27297032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08585644 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10365299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365301 - -
          Affymetrix HT MG-430 PM Array Plate 1431005_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1454648_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10365297 - -
          Mouse Genome 430 2.0 Array 1454648_PM_s_at - -
          Mouse Genome 430 2.0 Array 1454648_s_at - -
          Mouse Genome 430 2.0 Array 1431005_at - -
          Mouse Genome 430 2.0 Array 1431005_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 28109_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 28109_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000007506.mm.1 NM_026579.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007506.mm.1 NM_026579.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007506.mm.1 TC1000000940.mm.1 MTA TC
          Mouse Expression Set 430 1454648_s_at - -
          Mouse Expression Set 430 1454648_PM_s_at - -
          Mouse Expression Set 430 1431005_at - -
          Mouse Expression Set 430 1431005_PM_at - -
          Murine Genome U74v2 Set 160707_at - -
          Murine Genome U74v2 Set 160707_PM_at - -
          Mouse Genome 430A 2.0 Array 1454648_s_at - -
          Mouse Genome 430A 2.0 Array 1454648_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1454648_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1454648_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1431005_at - -
          Affymetrix HT Mouse Genome 430B Array 1431005_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 656023 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5360577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2707694 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2707694 - -
          mouse-6 v1.1 (Illumina) scl38624.3_492-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38624.3_492-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2707694 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2707694 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl38624.3_492-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline