Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01256301_m1
          See other FAM69C GE Assays ›
          Gene Symbol:
          Fam69c
          Gene Name:
          family with sequence similarity 69, member C
          Gene Aliases:
          B230399E16Rik
          Chromosome Location:
          Chr.18: 84718987 - 84741731 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01256301_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam69c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173770.4 NP_776131.1 4 - 5 1083 81
          GenBank mRNA AK046504.1 - 4 - 5 1084 81
          BC119040.1 - 2 - 3 671 8733789 81
          BC119042.1 - 2 - 3 671 8733791 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000886.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam69c

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31890269 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31890268 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31890267 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997479 NM_173770.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997479 AK046504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955120 NM_173770.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955120 AK046504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955120 BC119040.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955120 BC119042.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885969 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84385 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646847 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168545 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155017 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133447 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997479 NM_173770.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997479 AK046504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955120 NM_173770.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955120 AK046504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955120 BC119040.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955120 BC119042.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31890264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31890263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31890265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31890262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31890267 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03531648 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03531634 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133447 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155017 - -
          Whole Mouse Genome G4122A A_52_P680839 NM_173770.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P680839 AK046504.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111670 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133447 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155017 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14099754 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13913356 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13913365 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14099757 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31890269 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31890268 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31890267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31890432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13780292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13736456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14100456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14099757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13780288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13804201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13606547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13836483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13878447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13780303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14099759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14099756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13878454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14078766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13878446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13664987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13913353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31890301 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10457068 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457064 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457066 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457067 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457070 - -
          Affymetrix Mouse Gene 1.1 ST Array 10457063 - -
          Affymetrix Mouse Clariom S Array PSR1800006429.mm.1 NM_173770.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006429.mm.1 NM_173770.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006429.mm.1 TC1800000886.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800006431.mm.1 NM_173770.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006431.mm.1 NM_173770.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006431.mm.1 TC1800000886.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800006430.mm.1 NM_173770.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006430.mm.1 NM_173770.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006430.mm.1 TC1800000886.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 660646 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7100168 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100168 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230753 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230753 - -
          mouse-6 v1.1 (Illumina) scl51714.6.1_318-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100168 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51714.6.1_318-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100168 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230753 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230753 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl51714.6.1_318-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline