Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01257641_m1
          See other ZFP939 GE Assays ›
          Gene Symbol:
          Zfp939
          Gene Name:
          zinc finger protein 939
          Gene Aliases:
          9430025M13Rik
          Chromosome Location:
          Chr.7: 39449518 - 39477427 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01257641_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp939
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243021.1 NP_001229950.1 3 - 4 356 66
          GenBank mRNA AK034698.1 - 3 - 4 358 66

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144644 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121958 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965861 AK034698.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965861 AK162098.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03807072 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06171022 - -
          Whole Mouse Genome G4122A A_52_P3865 AK034698.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03807072 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06171022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875655 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552265 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552268 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552267 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552266 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552269 - -
          Affymetrix HT MG-430 PM Array Plate 1444092_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552264 - -
          Mouse Genome 430 2.0 Array 1444092_PM_at - -
          Mouse Genome 430 2.0 Array 1444092_at - -
          Mouse Expression Set 430 1444092_at - -
          Mouse Expression Set 430 1444092_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444092_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444092_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 752915 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646551 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2646551 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline